Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104968394-104968663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP4
Entrez Gene ID
837
Gene Name
caspase 4
Gene Aliases
ICE(rel)II, ICEREL-II, ICH-2, Mih1, Mih1/TX, TX
UniGene
Hs.138378
Gene Chromosome Location
Chr.11: 104942866-104968598 on Build GRCh38
Transcripts
NM_001225.3 XM_011543019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038563 c.7+2T>G p.? Chr.11: 104968517-104968517 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376476438 Chr.11: 104968587-104968587 on Build GRCh38
rs569135175 Chr.11: 104968629-104968629 on Build GRCh38
rs1014176490 Chr.11: 104968441-104968441 on Build GRCh38
rs56291433 Chr.11: 104968423-104968423 on Build GRCh38
rs996417755 Chr.11: 104968519-104968519 on Build GRCh38
rs754195610 Chr.11: 104968460-104968460 on Build GRCh38
rs907986884 Chr.11: 104968623-104968623 on Build GRCh38
rs755883291 Chr.11: 104968511-104968511 on Build GRCh38
rs762197509 Chr.11: 104968478-104968478 on Build GRCh38
rs772040846 Chr.11: 104968524-104968524 on Build GRCh38
rs376903172 Chr.11: 104968546-104968546 on Build GRCh38
rs56176061 Chr.11: 104968627-104968627 on Build GRCh38
rs193196983 Chr.11: 104968492-104968492 on Build GRCh38
rs745849708 Chr.11: 104968522-104968522 on Build GRCh38
rs752061641 Chr.11: 104968575-104968575 on Build GRCh38
rs764267939 Chr.11: 104968474-104968474 on Build GRCh38
rs750907867 Chr.11: 104968486-104968486 on Build GRCh38
rs751567166 Chr.11: 104968594-104968594 on Build GRCh38
rs200442223 Chr.11: 104968517-104968517 on Build GRCh38
rs377559194 Chr.11: 104968534-104968534 on Build GRCh38
rs145262923 Chr.11: 104968537-104968537 on Build GRCh38
rs765582795 Chr.11: 104968554-104968554 on Build GRCh38
rs748905548 Chr.11: 104968513-104968513 on Build GRCh38
rs1036945231 Chr.11: 104968425-104968425 on Build GRCh38
rs78953844 Chr.11: 104968531-104968531 on Build GRCh38
rs754899634 Chr.11: 104968613-104968613 on Build GRCh38
rs999882590 Chr.11: 104968525-104968525 on Build GRCh38
rs752227202 Chr.11: 104968496-104968496 on Build GRCh38
rs777588239 Chr.11: 104968512-104968512 on Build GRCh38
rs55803114 Chr.11: 104968426-104968426 on Build GRCh38
rs757003203 Chr.11: 104968515-104968515 on Build GRCh38
rs374669255 Chr.11: 104968543-104968543 on Build GRCh38
rs115796900 Chr.11: 104968592-104968592 on Build GRCh38
rs72978981 Chr.11: 104968573-104968573 on Build GRCh38
rs199666174 Chr.11: 104968540-104968540 on Build GRCh38
rs658560 Chr.11: 104968495-104968495 on Build GRCh38
rs773625877 Chr.11: 104968566-104968566 on Build GRCh38
rs773848270 Chr.11: 104968560-104968560 on Build GRCh38
rs765571945 Chr.11: 104968485-104968485 on Build GRCh38
rs916284048 Chr.11: 104968630-104968630 on Build GRCh38
rs762541912 Chr.11: 104968473-104968474 on Build GRCh38
rs760952282 Chr.11: 104968473-104968473 on Build GRCh38
rs532512892 Chr.11: 104968609-104968609 on Build GRCh38
rs761965905 Chr.11: 104968549-104968549 on Build GRCh38
rs573514402 Chr.11: 104968477-104968477 on Build GRCh38
rs1019226203 Chr.11: 104968432-104968433 on Build GRCh38
rs777531139 Chr.11: 104968530-104968530 on Build GRCh38
rs185020936 Chr.11: 104968599-104968599 on Build GRCh38
rs780981586 Chr.11: 104968625-104968625 on Build GRCh38
rs766056790 Chr.11: 104968543-104968544 on Build GRCh38

View Full Product Details