Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7483460-7483713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD163
Entrez Gene ID
9332
Gene Name
CD163 molecule
Gene Aliases
M130, MM130, SCARI1
UniGene
Hs.504641
Gene Chromosome Location
Chr.12: 7470816-7503818 on Build GRCh38
Transcripts
NM_004244.5 NM_203416.3 XM_005253528.4 XM_005253529.4 XM_017020231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4540191 c.2801G>A p.G934E Chr.12: 7483654-7483654 on Build GRCh38
3872519 c.2941G>A p.E981K Chr.12: 7483514-7483514 on Build GRCh38
6073554 c.2839G>T p.G947* Chr.12: 7483616-7483616 on Build GRCh38
7195867 c.2909T>G p.L970R Chr.12: 7483546-7483546 on Build GRCh38
7195868 c.3008T>G p.L1003R Chr.12: 7483546-7483546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768204023 Chr.12: 7483486-7483486 on Build GRCh38
rs776402823 Chr.12: 7483494-7483494 on Build GRCh38
rs1045655047 Chr.12: 7483488-7483488 on Build GRCh38
rs775082662 Chr.12: 7483485-7483485 on Build GRCh38
rs757152352 Chr.12: 7483617-7483617 on Build GRCh38
rs750247723 Chr.12: 7483625-7483625 on Build GRCh38
rs371406699 Chr.12: 7483572-7483572 on Build GRCh38
rs753885158 Chr.12: 7483613-7483613 on Build GRCh38
rs763870982 Chr.12: 7483571-7483571 on Build GRCh38
rs764142833 Chr.12: 7483574-7483574 on Build GRCh38
rs776200093 Chr.12: 7483499-7483499 on Build GRCh38
rs199560015 Chr.12: 7483641-7483641 on Build GRCh38
rs771413158 Chr.12: 7483518-7483518 on Build GRCh38
rs780582176 Chr.12: 7483669-7483669 on Build GRCh38
rs763374425 Chr.12: 7483511-7483511 on Build GRCh38
rs746721451 Chr.12: 7483635-7483635 on Build GRCh38
rs761565641 Chr.12: 7483626-7483626 on Build GRCh38
rs372790972 Chr.12: 7483557-7483557 on Build GRCh38
rs774603375 Chr.12: 7483527-7483527 on Build GRCh38
rs141756075 Chr.12: 7483564-7483564 on Build GRCh38
rs753020548 Chr.12: 7483563-7483563 on Build GRCh38
rs767965678 Chr.12: 7483548-7483548 on Build GRCh38
rs759943157 Chr.12: 7483531-7483531 on Build GRCh38
rs779801921 Chr.12: 7483631-7483631 on Build GRCh38
rs745473362 Chr.12: 7483618-7483618 on Build GRCh38
rs368897599 Chr.12: 7483566-7483566 on Build GRCh38
rs147938240 Chr.12: 7483675-7483675 on Build GRCh38
rs964444440 Chr.12: 7483634-7483634 on Build GRCh38

View Full Product Details