Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93436071-93436298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCR
Entrez Gene ID
799
Gene Name
calcitonin receptor
Gene Aliases
CRT, CT-R, CTR, CTR1
UniGene
Hs.489127
Gene Chromosome Location
Chr.7: 93424487-93574730 on Build GRCh38
Transcripts
NM_001164737.1 NM_001164738.1 NM_001742.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2864799 c.1107C>T p.S369S Chr.7: 93436096-93436096 on Build GRCh38
2864800 c.1005C>T p.S335S Chr.7: 93436096-93436096 on Build GRCh38
5665352 c.1106C>A p.S369Y Chr.7: 93436097-93436097 on Build GRCh38
747487 c.1039T>G p.F347V Chr.7: 93436164-93436164 on Build GRCh38
4520975 c.1087G>A p.E363K Chr.7: 93436116-93436116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556686258 Chr.7: 93436174-93436174 on Build GRCh38
rs762491371 Chr.7: 93436200-93436200 on Build GRCh38
rs369105382 Chr.7: 93436216-93436216 on Build GRCh38
rs775760519 Chr.7: 93436163-93436163 on Build GRCh38
rs747945692 Chr.7: 93436130-93436130 on Build GRCh38
rs747151239 Chr.7: 93436178-93436178 on Build GRCh38
rs200463057 Chr.7: 93436155-93436155 on Build GRCh38
rs371980647 Chr.7: 93436095-93436095 on Build GRCh38
rs113723720 Chr.7: 93436144-93436144 on Build GRCh38
rs148707949 Chr.7: 93436139-93436139 on Build GRCh38
rs765825396 Chr.7: 93436194-93436194 on Build GRCh38
rs765853136 Chr.7: 93436208-93436208 on Build GRCh38
rs545295161 Chr.7: 93436102-93436102 on Build GRCh38
rs140388547 Chr.7: 93436108-93436108 on Build GRCh38
rs780796922 Chr.7: 93436122-93436122 on Build GRCh38
rs751099849 Chr.7: 93436221-93436221 on Build GRCh38
rs201517548 Chr.7: 93436190-93436190 on Build GRCh38
rs1049867197 Chr.7: 93436237-93436237 on Build GRCh38
rs910140161 Chr.7: 93436140-93436140 on Build GRCh38
rs749894460 Chr.7: 93436093-93436093 on Build GRCh38
rs199570255 Chr.7: 93436162-93436162 on Build GRCh38
rs772892037 Chr.7: 93436126-93436126 on Build GRCh38
rs780371143 Chr.7: 93436172-93436184 on Build GRCh38
rs1031646573 Chr.7: 93436195-93436195 on Build GRCh38
rs901866737 Chr.7: 93436152-93436152 on Build GRCh38
rs575377286 Chr.7: 93436135-93436135 on Build GRCh38
rs372388923 Chr.7: 93436183-93436183 on Build GRCh38
rs1033749544 Chr.7: 93436116-93436116 on Build GRCh38
rs369253212 Chr.7: 93436166-93436166 on Build GRCh38
rs766301884 Chr.7: 93436103-93436103 on Build GRCh38
rs374733980 Chr.7: 93436196-93436196 on Build GRCh38
rs546143817 Chr.7: 93436182-93436182 on Build GRCh38
rs772087101 Chr.7: 93436153-93436153 on Build GRCh38
rs754776703 Chr.7: 93436115-93436115 on Build GRCh38

View Full Product Details