Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32203960-32204197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC28B
Entrez Gene ID
79140
Gene Name
coiled-coil domain containing 28B
Gene Aliases
-
UniGene
Hs.534482
Gene Chromosome Location
Chr.1: 32200386-32205390 on Build GRCh38
Transcripts
NM_001301011.1 NM_024296.4 XM_011542113.2 XM_017002306.1 XM_017002307.1 XM_017002308.1 XM_017002309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5435496 c.267T>C p.Y89Y Chr.1: 32203981-32203981 on Build GRCh38
5478478 c.264C>A p.V88V Chr.1: 32203978-32203978 on Build GRCh38
6622034 c.315C>T p.G105G Chr.1: 32204029-32204029 on Build GRCh38
319280 c.268G>C p.E90Q Chr.1: 32203982-32203982 on Build GRCh38
6622035 c.317G>A p.R106Q Chr.1: 32204031-32204031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751403021 Chr.1: 32204067-32204067 on Build GRCh38
rs952595550 Chr.1: 32204161-32204161 on Build GRCh38
rs200163249 Chr.1: 32203995-32203995 on Build GRCh38
rs753894609 Chr.1: 32204141-32204141 on Build GRCh38
rs759498949 Chr.1: 32204070-32204070 on Build GRCh38
rs761454975 Chr.1: 32204031-32204031 on Build GRCh38
rs777850558 Chr.1: 32203990-32203990 on Build GRCh38
rs561982752 Chr.1: 32204023-32204023 on Build GRCh38
rs988787948 Chr.1: 32204132-32204132 on Build GRCh38
rs202021004 Chr.1: 32204071-32204071 on Build GRCh38
rs764812524 Chr.1: 32204175-32204175 on Build GRCh38
rs1020083515 Chr.1: 32204116-32204116 on Build GRCh38
rs979217136 Chr.1: 32204055-32204055 on Build GRCh38
rs775797765 Chr.1: 32204013-32204013 on Build GRCh38
rs745773314 Chr.1: 32204028-32204028 on Build GRCh38
rs747436955 Chr.1: 32204178-32204178 on Build GRCh38
rs965493384 Chr.1: 32204131-32204131 on Build GRCh38
rs41263993 Chr.1: 32204044-32204044 on Build GRCh38
rs766209681 Chr.1: 32204062-32204062 on Build GRCh38
rs777877169 Chr.1: 32204084-32204107 on Build GRCh38
rs758635766 Chr.1: 32204169-32204169 on Build GRCh38
rs183850860 Chr.1: 32204137-32204137 on Build GRCh38
rs778998750 Chr.1: 32204153-32204153 on Build GRCh38
rs1001100101 Chr.1: 32203994-32203994 on Build GRCh38
rs913800131 Chr.1: 32204172-32204172 on Build GRCh38
rs749497454 Chr.1: 32204170-32204171 on Build GRCh38
rs772559410 Chr.1: 32204011-32204011 on Build GRCh38
rs776248241 Chr.1: 32204030-32204030 on Build GRCh38
rs762587976 Chr.1: 32204046-32204046 on Build GRCh38
rs757387251 Chr.1: 32204151-32204151 on Build GRCh38
rs750666303 Chr.1: 32204156-32204156 on Build GRCh38
rs749470747 Chr.1: 32203991-32203991 on Build GRCh38
rs772979728 Chr.1: 32204045-32204045 on Build GRCh38
rs756155701 Chr.1: 32204081-32204082 on Build GRCh38

View Full Product Details