Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 37142109-37142378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf42
Entrez Gene ID
65250
Gene Name
chromosome 5 open reading frame 42
Gene Aliases
Hug, JBTS17, OFD6
UniGene
Hs.586199
Gene Chromosome Location
Chr.5: 37063928-37249428 on Build GRCh38
Transcripts
NM_023073.3 XM_005248345.3 XM_005248346.3 XM_005248347.3 XM_005248349.3 XM_005248350.3 XM_006714491.3 XM_011514085.2 XM_011514086.2 XM_011514087.2 XM_011514088.2 XM_011514089.2 XM_011514090.2 XM_011514092.2 XM_011514094.2 XM_017009760.1 XM_017009761.1 XM_017009762.1 XM_017009763.1 XM_017009764.1 XM_017009765.1 XM_017009766.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616279 c.8407T>G p.F2803V Chr.5: 37142373-37142373 on Build GRCh38
4347843 c.5126G>A p.S1709N Chr.5: 37142348-37142348 on Build GRCh38
6616280 c.5101T>G p.F1701V Chr.5: 37142373-37142373 on Build GRCh38
4347842 c.8432G>A p.S2811N Chr.5: 37142348-37142348 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935322956 Chr.5: 37142248-37142248 on Build GRCh38
rs964459549 Chr.5: 37142185-37142185 on Build GRCh38
rs965839840 Chr.5: 37142157-37142157 on Build GRCh38
rs544915445 Chr.5: 37142317-37142317 on Build GRCh38
rs750310066 Chr.5: 37142330-37142330 on Build GRCh38
rs763705214 Chr.5: 37142315-37142315 on Build GRCh38
rs751260970 Chr.5: 37142316-37142316 on Build GRCh38
rs769286652 Chr.5: 37142289-37142289 on Build GRCh38
rs767191135 Chr.5: 37142160-37142160 on Build GRCh38
rs921855737 Chr.5: 37142165-37142165 on Build GRCh38
rs563494619 Chr.5: 37142319-37142319 on Build GRCh38
rs1014663981 Chr.5: 37142148-37142150 on Build GRCh38
rs1011615627 Chr.5: 37142220-37142220 on Build GRCh38
rs762550981 Chr.5: 37142307-37142307 on Build GRCh38
rs756020116 Chr.5: 37142339-37142339 on Build GRCh38
rs752341833 Chr.5: 37142348-37142348 on Build GRCh38
rs925241639 Chr.5: 37142245-37142245 on Build GRCh38
rs1008743245 Chr.5: 37142145-37142145 on Build GRCh38
rs754005437 Chr.5: 37142341-37142341 on Build GRCh38
rs774883520 Chr.5: 37142306-37142306 on Build GRCh38
rs1021630438 Chr.5: 37142300-37142300 on Build GRCh38
rs780136856 Chr.5: 37142340-37142340 on Build GRCh38
rs755153238 Chr.5: 37142345-37142345 on Build GRCh38

View Full Product Details