Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43977878-43978151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0B
Entrez Gene ID
533
Gene Name
ATPase H+ transporting V0 subunit b
Gene Aliases
ATP6F, HATPL, VMA16
UniGene
Hs.596514
Gene Chromosome Location
Chr.1: 43974648-43978300 on Build GRCh38
Transcripts
NM_001039457.2 NM_001294333.1 NM_004047.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3386191 c.592-1G>C p.? Chr.1: 43977980-43977980 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771663239 Chr.1: 43977933-43977933 on Build GRCh38
rs769384917 Chr.1: 43978026-43978027 on Build GRCh38
rs770142154 Chr.1: 43978044-43978044 on Build GRCh38
rs776315932 Chr.1: 43977969-43977970 on Build GRCh38
rs950320818 Chr.1: 43978036-43978036 on Build GRCh38
rs553041663 Chr.1: 43978067-43978067 on Build GRCh38
rs1053240003 Chr.1: 43977974-43977974 on Build GRCh38
rs946156343 Chr.1: 43978023-43978023 on Build GRCh38
rs888320735 Chr.1: 43978126-43978126 on Build GRCh38
rs763817422 Chr.1: 43977992-43977992 on Build GRCh38
rs374221606 Chr.1: 43978029-43978029 on Build GRCh38
rs35823146 Chr.1: 43978060-43978061 on Build GRCh38
rs145093221 Chr.1: 43977968-43977968 on Build GRCh38
rs1034577904 Chr.1: 43978097-43978097 on Build GRCh38
rs905842457 Chr.1: 43978065-43978065 on Build GRCh38
rs762232009 Chr.1: 43977943-43977943 on Build GRCh38
rs8494 Chr.1: 43978042-43978042 on Build GRCh38
rs573011195 Chr.1: 43978131-43978131 on Build GRCh38
rs752513305 Chr.1: 43977983-43977983 on Build GRCh38
rs933773868 Chr.1: 43977900-43977900 on Build GRCh38
rs985598898 Chr.1: 43977956-43977956 on Build GRCh38
rs139540839 Chr.1: 43977961-43977961 on Build GRCh38
rs537764074 Chr.1: 43977977-43977977 on Build GRCh38
rs776966051 Chr.1: 43977932-43977932 on Build GRCh38
rs751607900 Chr.1: 43977971-43977971 on Build GRCh38
rs879202782 Chr.1: 43978100-43978100 on Build GRCh38
rs767633842 Chr.1: 43977978-43977978 on Build GRCh38
rs112442160 Chr.1: 43977903-43977903 on Build GRCh38
rs747809739 Chr.1: 43978007-43978009 on Build GRCh38
rs528635093 Chr.1: 43977904-43977904 on Build GRCh38
rs763103960 Chr.1: 43977967-43977967 on Build GRCh38
rs756022638 Chr.1: 43977987-43977987 on Build GRCh38
rs1005346945 Chr.1: 43978128-43978128 on Build GRCh38
rs1027690058 Chr.1: 43978074-43978074 on Build GRCh38
rs14084 Chr.1: 43978026-43978026 on Build GRCh38
rs769968809 Chr.1: 43977952-43977952 on Build GRCh38
rs768220862 Chr.1: 43977967-43977970 on Build GRCh38
rs756866050 Chr.1: 43978014-43978014 on Build GRCh38
rs56842927 Chr.1: 43978037-43978037 on Build GRCh38
rs902606024 Chr.1: 43978112-43978125 on Build GRCh38
rs755472183 Chr.1: 43978028-43978028 on Build GRCh38
rs557265087 Chr.1: 43978011-43978011 on Build GRCh38
rs754773557 Chr.1: 43977941-43977941 on Build GRCh38
rs1001656927 Chr.1: 43978073-43978073 on Build GRCh38

View Full Product Details