Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139737869-139738130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11C
Entrez Gene ID
286410
Gene Name
ATPase phospholipid transporting 11C
Gene Aliases
ATPIG, ATPIQ
UniGene
Hs.88252
Gene Chromosome Location
Chr.X: 139726346-139933083 on Build GRCh38
Transcripts
NM_001010986.2 NM_173694.4 XM_017029439.1 XM_017029440.1 XM_017029441.1 XM_017029442.1 XM_017029443.1 XM_017029444.1 XM_017029445.1 XM_017029446.1 XM_017029447.1 XM_017029448.1 XM_017029449.1

SNP Detail

SNP ID Chromosome Location
rs774030771 Chr.X: 139737892-139737892 on Build GRCh38
rs770317627 Chr.X: 139738089-139738089 on Build GRCh38
rs890191882 Chr.X: 139738013-139738013 on Build GRCh38
rs746282132 Chr.X: 139738087-139738087 on Build GRCh38
rs201858795 Chr.X: 139737909-139737909 on Build GRCh38
rs750271759 Chr.X: 139737929-139737929 on Build GRCh38
rs748765155 Chr.X: 139738091-139738091 on Build GRCh38
rs768185582 Chr.X: 139738093-139738093 on Build GRCh38
rs868765474 Chr.X: 139738031-139738031 on Build GRCh38
rs1000008980 Chr.X: 139737974-139737975 on Build GRCh38
rs769874368 Chr.X: 139737923-139737925 on Build GRCh38
rs142741321 Chr.X: 139737987-139737987 on Build GRCh38
rs374132692 Chr.X: 139737938-139737938 on Build GRCh38
rs747268235 Chr.X: 139738000-139738000 on Build GRCh38
rs780587311 Chr.X: 139738090-139738090 on Build GRCh38
rs1050542132 Chr.X: 139737988-139737988 on Build GRCh38
rs766294642 Chr.X: 139737946-139737946 on Build GRCh38
rs781621410 Chr.X: 139738054-139738054 on Build GRCh38
rs778114321 Chr.X: 139737990-139737990 on Build GRCh38
rs770459250 Chr.X: 139738084-139738085 on Build GRCh38
rs757503094 Chr.X: 139738009-139738009 on Build GRCh38
rs1009101682 Chr.X: 139738036-139738036 on Build GRCh38
rs781505372 Chr.X: 139737905-139737905 on Build GRCh38
rs371297467 Chr.X: 139737918-139737918 on Build GRCh38

View Full Product Details