Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60456828-60457057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf39
Entrez Gene ID
317761
Gene Name
chromosome 14 open reading frame 39
Gene Aliases
Six6os1
UniGene
Hs.335754
Gene Chromosome Location
Chr.14: 60435954-60499773 on Build GRCh38
Transcripts
NM_174978.2 XM_011536690.2 XM_011536703.2 XM_017021247.1 XM_017021248.1 XM_017021249.1 XM_017021250.1 XM_017021251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4462652 c.1255C>T p.P419S Chr.14: 60457020-60457020 on Build GRCh38
222278 c.1349C>T p.P450L Chr.14: 60456926-60456926 on Build GRCh38
3386608 c.1219G>T p.D407Y Chr.14: 60457056-60457056 on Build GRCh38
4464806 c.1353C>T p.F451F Chr.14: 60456922-60456922 on Build GRCh38
4051570 c.1263G>A p.T421T Chr.14: 60457012-60457012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745638751 Chr.14: 60456933-60456933 on Build GRCh38
rs759887326 Chr.14: 60456973-60456973 on Build GRCh38
rs778017059 Chr.14: 60456926-60456926 on Build GRCh38
rs764734906 Chr.14: 60456964-60456964 on Build GRCh38
rs149583331 Chr.14: 60456999-60456999 on Build GRCh38
rs116107793 Chr.14: 60456940-60456940 on Build GRCh38
rs766802395 Chr.14: 60456882-60456882 on Build GRCh38
rs376214220 Chr.14: 60457018-60457018 on Build GRCh38
rs779000535 Chr.14: 60457000-60457000 on Build GRCh38
rs753179285 Chr.14: 60456985-60456985 on Build GRCh38
rs926328665 Chr.14: 60456936-60456936 on Build GRCh38
rs114644048 Chr.14: 60456966-60456966 on Build GRCh38
rs35442613 Chr.14: 60456992-60456992 on Build GRCh38
rs147627198 Chr.14: 60456921-60456921 on Build GRCh38
rs761535244 Chr.14: 60456962-60456962 on Build GRCh38
rs776870689 Chr.14: 60456925-60456926 on Build GRCh38
rs751800166 Chr.14: 60456896-60456896 on Build GRCh38
rs372630104 Chr.14: 60456953-60456953 on Build GRCh38
rs375791877 Chr.14: 60456866-60456866 on Build GRCh38
rs772204669 Chr.14: 60456938-60456938 on Build GRCh38
rs772705811 Chr.14: 60457025-60457025 on Build GRCh38
rs1008943486 Chr.14: 60456868-60456868 on Build GRCh38
rs776214261 Chr.14: 60457013-60457013 on Build GRCh38
rs749546937 Chr.14: 60456931-60456931 on Build GRCh38
rs775967405 Chr.14: 60456957-60456957 on Build GRCh38
rs77727318 Chr.14: 60456978-60456978 on Build GRCh38
rs764173083 Chr.14: 60456894-60456895 on Build GRCh38
rs200992791 Chr.14: 60456922-60456922 on Build GRCh38
rs138071695 Chr.14: 60456932-60456932 on Build GRCh38
rs758248760 Chr.14: 60457005-60457005 on Build GRCh38
rs562941948 Chr.14: 60456925-60456925 on Build GRCh38
rs115702062 Chr.14: 60456897-60456897 on Build GRCh38
rs148165852 Chr.14: 60457016-60457016 on Build GRCh38
rs745717405 Chr.14: 60457002-60457002 on Build GRCh38
rs779949517 Chr.14: 60457009-60457009 on Build GRCh38
rs756564448 Chr.14: 60456987-60456987 on Build GRCh38
rs1018950505 Chr.14: 60456874-60456874 on Build GRCh38
rs746977921 Chr.14: 60457012-60457012 on Build GRCh38
rs746569031 Chr.14: 60456943-60456943 on Build GRCh38
rs149339653 Chr.14: 60456996-60456996 on Build GRCh38

View Full Product Details