Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101405795-101406054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL1
Entrez Gene ID
400
Gene Name
ADP ribosylation factor like GTPase 1
Gene Aliases
ARFL1
UniGene
Hs.372616
Gene Chromosome Location
Chr.12: 101393120-101407820 on Build GRCh38
Transcripts
NM_001177.5 NM_001301068.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5777608 c.38T>G p.F13C Chr.12: 101405948-101405948 on Build GRCh38
4486006 c.29C>T p.S10F Chr.12: 101405957-101405957 on Build GRCh38
5636677 c.94A>T p.T32S Chr.12: 101405892-101405892 on Build GRCh38
2063371 c.47G>A p.R16Q Chr.12: 101405939-101405939 on Build GRCh38
6531646 c.133A>G p.T45A Chr.12: 101405853-101405853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748657017 Chr.12: 101406012-101406013 on Build GRCh38
rs768928432 Chr.12: 101406007-101406007 on Build GRCh38
rs199647502 Chr.12: 101405962-101405962 on Build GRCh38
rs779810772 Chr.12: 101405829-101405829 on Build GRCh38
rs766854112 Chr.12: 101405835-101405835 on Build GRCh38
rs374082637 Chr.12: 101405990-101405990 on Build GRCh38
rs756021715 Chr.12: 101405950-101405950 on Build GRCh38
rs866509393 Chr.12: 101405846-101405846 on Build GRCh38
rs889507227 Chr.12: 101405943-101405943 on Build GRCh38
rs747215626 Chr.12: 101406003-101406003 on Build GRCh38
rs745925566 Chr.12: 101405976-101405976 on Build GRCh38
rs1052656106 Chr.12: 101405833-101405833 on Build GRCh38
rs367987094 Chr.12: 101405992-101405992 on Build GRCh38
rs997604444 Chr.12: 101405872-101405872 on Build GRCh38
rs1050813719 Chr.12: 101405899-101405899 on Build GRCh38
rs747900626 Chr.12: 101405889-101405889 on Build GRCh38
rs962695080 Chr.12: 101406009-101406009 on Build GRCh38
rs776910648 Chr.12: 101406011-101406011 on Build GRCh38
rs373636760 Chr.12: 101405836-101405836 on Build GRCh38

View Full Product Details