Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48447007-48447232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRIP
Entrez Gene ID
84126
Gene Name
ATR interacting protein
Gene Aliases
-
UniGene
Hs.694840
Gene Chromosome Location
Chr.3: 48446710-48465655 on Build GRCh38
Transcripts
NM_001271022.1 NM_001271023.1 NM_032166.3 NM_130384.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4516624 c.209_210CC>TT p.A70V Chr.3: 48447054-48447055 on Build GRCh38
5016001 c.186_189delGCTT p.L63fs*8 Chr.3: 48447031-48447034 on Build GRCh38
1683065 c.190G>T p.D64Y Chr.3: 48447035-48447035 on Build GRCh38
4664604 c.235C>T p.R79W Chr.3: 48447080-48447080 on Build GRCh38
5991737 c.215_220delGCCAAT p.Q73_C74delQC Chr.3: 48447060-48447065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370892766 Chr.3: 48447108-48447108 on Build GRCh38
rs767653932 Chr.3: 48447085-48447085 on Build GRCh38
rs759584758 Chr.3: 48447084-48447084 on Build GRCh38
rs147301531 Chr.3: 48447187-48447187 on Build GRCh38
rs758297238 Chr.3: 48447066-48447066 on Build GRCh38
rs947277468 Chr.3: 48447051-48447051 on Build GRCh38
rs555951256 Chr.3: 48447135-48447135 on Build GRCh38
rs770535111 Chr.3: 48447162-48447162 on Build GRCh38
rs943957532 Chr.3: 48447086-48447086 on Build GRCh38
rs745361845 Chr.3: 48447044-48447044 on Build GRCh38
rs763731046 Chr.3: 48447123-48447123 on Build GRCh38
rs772976477 Chr.3: 48447041-48447041 on Build GRCh38
rs753699864 Chr.3: 48447127-48447127 on Build GRCh38
rs757094875 Chr.3: 48447135-48447135 on Build GRCh38
rs148174212 Chr.3: 48447062-48447062 on Build GRCh38
rs746715933 Chr.3: 48447053-48447053 on Build GRCh38
rs374884213 Chr.3: 48447137-48447137 on Build GRCh38
rs375576948 Chr.3: 48447134-48447134 on Build GRCh38
rs757223539 Chr.3: 48447039-48447039 on Build GRCh38
rs760902972 Chr.3: 48447116-48447116 on Build GRCh38
rs750450647 Chr.3: 48447138-48447138 on Build GRCh38
rs1000190732 Chr.3: 48447040-48447040 on Build GRCh38
rs900301715 Chr.3: 48447070-48447070 on Build GRCh38
rs1014568056 Chr.3: 48447174-48447174 on Build GRCh38
rs748988992 Chr.3: 48447150-48447150 on Build GRCh38
rs1008794343 Chr.3: 48447045-48447045 on Build GRCh38
rs144998094 Chr.3: 48447064-48447064 on Build GRCh38
rs779847347 Chr.3: 48447049-48447049 on Build GRCh38
rs777683743 Chr.3: 48447026-48447026 on Build GRCh38
rs770223988 Chr.3: 48447056-48447056 on Build GRCh38
rs778246445 Chr.3: 48447043-48447043 on Build GRCh38
rs771487037 Chr.3: 48447069-48447069 on Build GRCh38
rs529538750 Chr.3: 48447038-48447038 on Build GRCh38
rs551289572 Chr.3: 48447048-48447048 on Build GRCh38
rs372906254 Chr.3: 48447074-48447074 on Build GRCh38
rs757967012 Chr.3: 48447141-48447141 on Build GRCh38
rs996417186 Chr.3: 48447092-48447092 on Build GRCh38

View Full Product Details