Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70158280-70158505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR27
Entrez Gene ID
401137
Gene Name
proline rich 27
Gene Aliases
C4orf40
UniGene
Hs.518795
Gene Chromosome Location
Chr.4: 70154187-70166609 on Build GRCh38
Transcripts
NM_214711.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1166253 c.161G>A p.R54H Chr.4: 70158413-70158413 on Build GRCh38
4585154 c.191G>C p.G64A Chr.4: 70158443-70158443 on Build GRCh38
6885336 c.221C>T p.S74L Chr.4: 70158473-70158473 on Build GRCh38
1057037 c.160C>T p.R54C Chr.4: 70158412-70158412 on Build GRCh38
4569561 c.184T>C p.Y62H Chr.4: 70158436-70158436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767283531 Chr.4: 70158374-70158374 on Build GRCh38
rs767178631 Chr.4: 70158458-70158458 on Build GRCh38
rs781456072 Chr.4: 70158427-70158427 on Build GRCh38
rs897470167 Chr.4: 70158356-70158356 on Build GRCh38
rs1612460 Chr.4: 70158382-70158382 on Build GRCh38
rs145691438 Chr.4: 70158415-70158415 on Build GRCh38
rs762485479 Chr.4: 70158347-70158347 on Build GRCh38
rs747195575 Chr.4: 70158309-70158309 on Build GRCh38
rs1717577 Chr.4: 70158400-70158400 on Build GRCh38
rs147899116 Chr.4: 70158412-70158412 on Build GRCh38
rs141586629 Chr.4: 70158454-70158454 on Build GRCh38
rs770259538 Chr.4: 70158342-70158342 on Build GRCh38
rs375338764 Chr.4: 70158378-70158378 on Build GRCh38
rs746412473 Chr.4: 70158340-70158340 on Build GRCh38
rs766217684 Chr.4: 70158473-70158473 on Build GRCh38
rs1015593919 Chr.4: 70158349-70158349 on Build GRCh38
rs763532150 Chr.4: 70158364-70158364 on Build GRCh38
rs372651799 Chr.4: 70158438-70158438 on Build GRCh38
rs267600231 Chr.4: 70158373-70158373 on Build GRCh38
rs149805235 Chr.4: 70158411-70158411 on Build GRCh38
rs888020016 Chr.4: 70158341-70158341 on Build GRCh38
rs746249276 Chr.4: 70158436-70158436 on Build GRCh38
rs768111718 Chr.4: 70158453-70158453 on Build GRCh38
rs971810835 Chr.4: 70158474-70158474 on Build GRCh38
rs777142041 Chr.4: 70158446-70158447 on Build GRCh38
rs760579021 Chr.4: 70158468-70158468 on Build GRCh38
rs371578811 Chr.4: 70158346-70158346 on Build GRCh38
rs777047004 Chr.4: 70158329-70158329 on Build GRCh38
rs546416421 Chr.4: 70158440-70158440 on Build GRCh38
rs749742607 Chr.4: 70158446-70158446 on Build GRCh38
rs770829246 Chr.4: 70158413-70158413 on Build GRCh38
rs576421826 Chr.4: 70158414-70158414 on Build GRCh38
rs758566145 Chr.4: 70158403-70158403 on Build GRCh38
rs376628196 Chr.4: 70158455-70158455 on Build GRCh38
rs544369111 Chr.4: 70158327-70158327 on Build GRCh38
rs190661582 Chr.4: 70158385-70158385 on Build GRCh38
rs772951766 Chr.4: 70158466-70158466 on Build GRCh38

View Full Product Details