Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110900325-110900586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBIP1
Entrez Gene ID
92482
Gene Name
BBSome interacting protein 1
Gene Aliases
BBIP10, BBS18, NCRNA00081, bA348N5.3
UniGene
Hs.232543
Gene Chromosome Location
Chr.10: 110898730-110919366 on Build GRCh38
Transcripts
NM_001195304.1 NM_001195305.1 NM_001195306.1 NM_001195307.1 NM_001243783.1

SNP Detail

SNP ID Chromosome Location
rs924008623 Chr.10: 110900509-110900509 on Build GRCh38
rs11548767 Chr.10: 110900502-110900502 on Build GRCh38
rs565980586 Chr.10: 110900497-110900497 on Build GRCh38
rs912754275 Chr.10: 110900523-110900523 on Build GRCh38
rs982682708 Chr.10: 110900469-110900469 on Build GRCh38
rs753007436 Chr.10: 110900499-110900499 on Build GRCh38
rs1024829955 Chr.10: 110900350-110900350 on Build GRCh38
rs769560942 Chr.10: 110900522-110900522 on Build GRCh38
rs767511014 Chr.10: 110900489-110900489 on Build GRCh38
rs368712453 Chr.10: 110900553-110900553 on Build GRCh38
rs781040735 Chr.10: 110900376-110900376 on Build GRCh38
rs879046499 Chr.10: 110900353-110900353 on Build GRCh38
rs981576340 Chr.10: 110900377-110900377 on Build GRCh38
rs991079395 Chr.10: 110900488-110900489 on Build GRCh38
rs760228426 Chr.10: 110900519-110900519 on Build GRCh38
rs768526182 Chr.10: 110900473-110900473 on Build GRCh38
rs547687357 Chr.10: 110900457-110900457 on Build GRCh38
rs777989008 Chr.10: 110900361-110900361 on Build GRCh38
rs914790477 Chr.10: 110900474-110900474 on Build GRCh38
rs762602315 Chr.10: 110900426-110900426 on Build GRCh38
rs548345581 Chr.10: 110900537-110900537 on Build GRCh38
rs761751086 Chr.10: 110900481-110900481 on Build GRCh38
rs990110859 Chr.10: 110900471-110900471 on Build GRCh38
rs950517725 Chr.10: 110900458-110900458 on Build GRCh38
rs774089309 Chr.10: 110900479-110900479 on Build GRCh38
rs767183949 Chr.10: 110900407-110900407 on Build GRCh38
rs890296233 Chr.10: 110900552-110900552 on Build GRCh38
rs515726134 Chr.10: 110900466-110900466 on Build GRCh38
rs776957044 Chr.10: 110900413-110900413 on Build GRCh38
rs1049860215 Chr.10: 110900512-110900512 on Build GRCh38
rs924597328 Chr.10: 110900385-110900385 on Build GRCh38
rs532149239 Chr.10: 110900406-110900406 on Build GRCh38
rs1026036041 Chr.10: 110900457-110900458 on Build GRCh38

View Full Product Details