Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154352588-154352849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
UniGene
Hs.195464
Gene Chromosome Location
Chr.X: 154348529-154374638 on Build GRCh38
Transcripts
NM_001110556.1 NM_001456.3 XM_011531127.1 XM_011531128.1 XM_011531129.1 XM_011531130.1 XM_011531131.1

SNP Detail

SNP ID Chromosome Location
rs782587942 Chr.X: 154352729-154352729 on Build GRCh38
rs782161469 Chr.X: 154352774-154352774 on Build GRCh38
rs782655948 Chr.X: 154352700-154352700 on Build GRCh38
rs375205247 Chr.X: 154352801-154352801 on Build GRCh38
rs201932105 Chr.X: 154352609-154352609 on Build GRCh38
rs201396725 Chr.X: 154352661-154352661 on Build GRCh38
rs201030213 Chr.X: 154352727-154352727 on Build GRCh38
rs782107005 Chr.X: 154352788-154352788 on Build GRCh38
rs781978328 Chr.X: 154352753-154352753 on Build GRCh38
rs1028791955 Chr.X: 154352772-154352772 on Build GRCh38
rs781923813 Chr.X: 154352767-154352767 on Build GRCh38
rs200524526 Chr.X: 154352643-154352643 on Build GRCh38
rs781999359 Chr.X: 154352779-154352779 on Build GRCh38
rs781945322 Chr.X: 154352627-154352627 on Build GRCh38
rs782467286 Chr.X: 154352674-154352674 on Build GRCh38
rs782289211 Chr.X: 154352705-154352705 on Build GRCh38
rs782316013 Chr.X: 154352620-154352620 on Build GRCh38
rs782096496 Chr.X: 154352636-154352636 on Build GRCh38
rs782142395 Chr.X: 154352646-154352646 on Build GRCh38
rs786205201 Chr.X: 154352795-154352796 on Build GRCh38
rs373398860 Chr.X: 154352755-154352755 on Build GRCh38
rs782691221 Chr.X: 154352827-154352827 on Build GRCh38
rs782260496 Chr.X: 154352759-154352759 on Build GRCh38
rs782402511 Chr.X: 154352615-154352615 on Build GRCh38
rs782400832 Chr.X: 154352775-154352775 on Build GRCh38
rs994764879 Chr.X: 154352693-154352693 on Build GRCh38
rs863223634 Chr.X: 154352667-154352668 on Build GRCh38
rs1057518479 Chr.X: 154352813-154352813 on Build GRCh38
rs369979360 Chr.X: 154352752-154352752 on Build GRCh38
rs782203408 Chr.X: 154352735-154352735 on Build GRCh38
rs782061936 Chr.X: 154352820-154352820 on Build GRCh38
rs1004981975 Chr.X: 154352647-154352647 on Build GRCh38
rs782022901 Chr.X: 154352613-154352613 on Build GRCh38
rs782740311 Chr.X: 154352800-154352800 on Build GRCh38
rs201773416 Chr.X: 154352699-154352699 on Build GRCh38
rs782405240 Chr.X: 154352765-154352765 on Build GRCh38
rs782761341 Chr.X: 154352640-154352640 on Build GRCh38
rs200832880 Chr.X: 154352619-154352619 on Build GRCh38
rs953354307 Chr.X: 154352776-154352776 on Build GRCh38

View Full Product Details