Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3667276-3667541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA10
Entrez Gene ID
57053
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
Gene Aliases
-
UniGene
Hs.732806
Gene Chromosome Location
Chr.11: 3665587-3673208 on Build GRCh38
Transcripts
NM_001303034.1 NM_001303035.1 NM_020402.3 XM_011520234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431518 c.685A>G p.T229A Chr.11: 3667442-3667442 on Build GRCh38
4963690 c.742G>A p.V248M Chr.11: 3667385-3667385 on Build GRCh38
926771 c.740G>A p.C247Y Chr.11: 3667387-3667387 on Build GRCh38
926772 c.718G>A p.V240M Chr.11: 3667409-3667409 on Build GRCh38
7287133 c.830C>T p.A277V Chr.11: 3667297-3667297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781223689 Chr.11: 3667332-3667332 on Build GRCh38
rs376404549 Chr.11: 3667412-3667412 on Build GRCh38
rs776814904 Chr.11: 3667482-3667482 on Build GRCh38
rs760737901 Chr.11: 3667416-3667416 on Build GRCh38
rs755960899 Chr.11: 3667346-3667346 on Build GRCh38
rs777354719 Chr.11: 3667426-3667426 on Build GRCh38
rs777343278 Chr.11: 3667394-3667394 on Build GRCh38
rs753813237 Chr.11: 3667388-3667388 on Build GRCh38
rs763249970 Chr.11: 3667315-3667315 on Build GRCh38
rs773474519 Chr.11: 3667415-3667415 on Build GRCh38
rs373143429 Chr.11: 3667454-3667454 on Build GRCh38
rs765656579 Chr.11: 3667297-3667297 on Build GRCh38
rs780569791 Chr.11: 3667503-3667503 on Build GRCh38
rs754915280 Chr.11: 3667420-3667420 on Build GRCh38
rs557047608 Chr.11: 3667430-3667430 on Build GRCh38
rs370718990 Chr.11: 3667308-3667308 on Build GRCh38
rs772373582 Chr.11: 3667511-3667511 on Build GRCh38
rs370949315 Chr.11: 3667418-3667418 on Build GRCh38
rs201153030 Chr.11: 3667513-3667513 on Build GRCh38
rs200192816 Chr.11: 3667485-3667485 on Build GRCh38
rs747774482 Chr.11: 3667362-3667362 on Build GRCh38
rs140742217 Chr.11: 3667331-3667331 on Build GRCh38
rs201163422 Chr.11: 3667479-3667479 on Build GRCh38
rs756764123 Chr.11: 3667429-3667429 on Build GRCh38
rs758085196 Chr.11: 3667392-3667392 on Build GRCh38
rs758459180 Chr.11: 3667486-3667512 on Build GRCh38
rs762468164 Chr.11: 3667508-3667508 on Build GRCh38
rs779466362 Chr.11: 3667507-3667507 on Build GRCh38
rs577106168 Chr.11: 3667498-3667498 on Build GRCh38
rs546692965 Chr.11: 3667419-3667419 on Build GRCh38
rs746558230 Chr.11: 3667396-3667396 on Build GRCh38
rs771119230 Chr.11: 3667518-3667518 on Build GRCh38
rs752898080 Chr.11: 3667450-3667451 on Build GRCh38
rs762696179 Chr.11: 3667491-3667491 on Build GRCh38
rs751774593 Chr.11: 3667321-3667321 on Build GRCh38
rs778830088 Chr.11: 3667390-3667390 on Build GRCh38
rs746641185 Chr.11: 3667427-3667427 on Build GRCh38
rs772484008 Chr.11: 3667457-3667457 on Build GRCh38
rs748982511 Chr.11: 3667353-3667353 on Build GRCh38
rs866721767 Chr.11: 3667505-3667505 on Build GRCh38
rs764409011 Chr.11: 3667377-3667377 on Build GRCh38
rs760932993 Chr.11: 3667467-3667467 on Build GRCh38
rs776864644 Chr.11: 3667519-3667519 on Build GRCh38
rs763375115 Chr.11: 3667375-3667375 on Build GRCh38
rs759872210 Chr.11: 3667422-3667424 on Build GRCh38
rs568176743 Chr.11: 3667421-3667421 on Build GRCh38
rs770470937 Chr.11: 3667397-3667397 on Build GRCh38
rs747228623 Chr.11: 3667514-3667514 on Build GRCh38
rs978017903 Chr.11: 3667410-3667410 on Build GRCh38
rs535473918 Chr.11: 3667424-3667424 on Build GRCh38
rs896866390 Chr.11: 3667458-3667458 on Build GRCh38
rs759550632 Chr.11: 3667483-3667483 on Build GRCh38
rs769081824 Chr.11: 3667448-3667448 on Build GRCh38
rs756891520 Chr.11: 3667501-3667501 on Build GRCh38
rs140976405 Chr.11: 3667366-3667366 on Build GRCh38
rs774655790 Chr.11: 3667409-3667409 on Build GRCh38
rs146548502 Chr.11: 3667359-3667359 on Build GRCh38
rs144182744 Chr.11: 3667348-3667348 on Build GRCh38
rs751827575 Chr.11: 3667380-3667380 on Build GRCh38
rs2231543 Chr.11: 3667363-3667363 on Build GRCh38
rs770133301 Chr.11: 3667374-3667374 on Build GRCh38
rs909765043 Chr.11: 3667388-3667411 on Build GRCh38
rs756087818 Chr.11: 3667386-3667386 on Build GRCh38
rs747827541 Chr.11: 3667391-3667391 on Build GRCh38
rs576316543 Chr.11: 3667431-3667431 on Build GRCh38
rs776973836 Chr.11: 3667309-3667328 on Build GRCh38
rs570947917 Chr.11: 3667411-3667411 on Build GRCh38
rs764357729 Chr.11: 3667320-3667320 on Build GRCh38
rs888647871 Chr.11: 3667364-3667364 on Build GRCh38
rs761971862 Chr.11: 3667382-3667382 on Build GRCh38
rs369074831 Chr.11: 3667441-3667441 on Build GRCh38
rs754864013 Chr.11: 3667389-3667389 on Build GRCh38
rs773351290 Chr.11: 3667466-3667466 on Build GRCh38
rs955026093 Chr.11: 3667432-3667432 on Build GRCh38
rs201720948 Chr.11: 3667314-3667314 on Build GRCh38
rs763924776 Chr.11: 3667496-3667496 on Build GRCh38
rs1048686864 Chr.11: 3667490-3667490 on Build GRCh38
rs759939729 Chr.11: 3667296-3667296 on Build GRCh38
rs373527871 Chr.11: 3667406-3667406 on Build GRCh38
rs765438697 Chr.11: 3667443-3667445 on Build GRCh38
rs775899850 Chr.11: 3667401-3667401 on Build GRCh38
rs774706922 Chr.11: 3667452-3667452 on Build GRCh38
rs750393817 Chr.11: 3667336-3667336 on Build GRCh38
rs748626494 Chr.11: 3667510-3667510 on Build GRCh38
rs148870170 Chr.11: 3667422-3667422 on Build GRCh38
rs201793421 Chr.11: 3667504-3667504 on Build GRCh38
rs2231542 Chr.11: 3667385-3667385 on Build GRCh38
rs138838925 Chr.11: 3667387-3667387 on Build GRCh38

View Full Product Details