Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45544579-45544830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH6B
Entrez Gene ID
220081
Gene Name
glutamate rich 6B
Gene Aliases
FAM194B
UniGene
Hs.668747
Gene Chromosome Location
Chr.13: 45541203-45615739 on Build GRCh38
Transcripts
NM_182542.2 XM_011534965.2 XM_017020418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1367023 c.1823T>C p.F608S Chr.13: 45544809-45544809 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs949946728 Chr.13: 45544774-45544774 on Build GRCh38
rs773951795 Chr.13: 45544736-45544736 on Build GRCh38
rs1024021876 Chr.13: 45544648-45544648 on Build GRCh38
rs761424106 Chr.13: 45544754-45544754 on Build GRCh38
rs370891897 Chr.13: 45544714-45544714 on Build GRCh38
rs547200532 Chr.13: 45544640-45544640 on Build GRCh38
rs925820080 Chr.13: 45544720-45544720 on Build GRCh38
rs867618281 Chr.13: 45544792-45544792 on Build GRCh38
rs957304383 Chr.13: 45544747-45544747 on Build GRCh38
rs767496773 Chr.13: 45544773-45544773 on Build GRCh38
rs750394584 Chr.13: 45544785-45544785 on Build GRCh38
rs1031063997 Chr.13: 45544697-45544697 on Build GRCh38
rs534979721 Chr.13: 45544631-45544631 on Build GRCh38
rs967151024 Chr.13: 45544702-45544702 on Build GRCh38
rs927107227 Chr.13: 45544803-45544803 on Build GRCh38
rs369400264 Chr.13: 45544663-45544663 on Build GRCh38
rs568546736 Chr.13: 45544641-45544641 on Build GRCh38

View Full Product Details