Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158972934-158973195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf45
Entrez Gene ID
152940
Gene Name
chromosome 4 open reading frame 45
Gene Aliases
-
UniGene
Hs.415576
Gene Chromosome Location
Chr.4: 158893532-159082883 on Build GRCh38
Transcripts
NM_152543.2 XM_011531671.2 XM_011531672.2 XM_017007808.1 XM_017007809.1 XM_017007810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616161 c.263A>G p.N88S Chr.4: 158973113-158973113 on Build GRCh38
5091165 c.254A>C p.N85T Chr.4: 158973122-158973122 on Build GRCh38
5159103 c.297+7C>T p.? Chr.4: 158973072-158973072 on Build GRCh38
3334828 c.257T>G p.F86C Chr.4: 158973119-158973119 on Build GRCh38
3601491 c.244C>T p.P82S Chr.4: 158973132-158973132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911527556 Chr.4: 158973109-158973109 on Build GRCh38
rs764138731 Chr.4: 158973039-158973039 on Build GRCh38
rs768857202 Chr.4: 158973136-158973136 on Build GRCh38
rs1033342192 Chr.4: 158972980-158972980 on Build GRCh38
rs752348666 Chr.4: 158973059-158973059 on Build GRCh38
rs557543735 Chr.4: 158972985-158972985 on Build GRCh38
rs763751751 Chr.4: 158973163-158973163 on Build GRCh38
rs779356187 Chr.4: 158973086-158973086 on Build GRCh38
rs193096054 Chr.4: 158973089-158973089 on Build GRCh38
rs753948728 Chr.4: 158973040-158973040 on Build GRCh38
rs777148354 Chr.4: 158973073-158973073 on Build GRCh38
rs774477361 Chr.4: 158973154-158973154 on Build GRCh38
rs767803611 Chr.4: 158973156-158973156 on Build GRCh38
rs1820511 Chr.4: 158973047-158973047 on Build GRCh38
rs141091800 Chr.4: 158972995-158972995 on Build GRCh38
rs114910865 Chr.4: 158972991-158972991 on Build GRCh38
rs982164913 Chr.4: 158973022-158973022 on Build GRCh38
rs184637066 Chr.4: 158973157-158973157 on Build GRCh38
rs926703195 Chr.4: 158973023-158973023 on Build GRCh38
rs374884319 Chr.4: 158973110-158973110 on Build GRCh38
rs754858076 Chr.4: 158973050-158973050 on Build GRCh38
rs775913788 Chr.4: 158973101-158973101 on Build GRCh38
rs996754578 Chr.4: 158972996-158972996 on Build GRCh38
rs755783714 Chr.4: 158973060-158973060 on Build GRCh38
rs371994683 Chr.4: 158973160-158973160 on Build GRCh38
rs762990905 Chr.4: 158973042-158973042 on Build GRCh38
rs756458437 Chr.4: 158973139-158973142 on Build GRCh38
rs767321684 Chr.4: 158973053-158973053 on Build GRCh38
rs776589684 Chr.4: 158973144-158973144 on Build GRCh38
rs886490919 Chr.4: 158972977-158972977 on Build GRCh38
rs1007932156 Chr.4: 158973041-158973041 on Build GRCh38
rs1028268996 Chr.4: 158973000-158973000 on Build GRCh38
rs766611983 Chr.4: 158973052-158973052 on Build GRCh38
rs662473 Chr.4: 158973153-158973153 on Build GRCh38
rs619128 Chr.4: 158973098-158973098 on Build GRCh38
rs753380148 Chr.4: 158973164-158973164 on Build GRCh38
rs761936920 Chr.4: 158973150-158973150 on Build GRCh38
rs62351583 Chr.4: 158973159-158973159 on Build GRCh38
rs535188981 Chr.4: 158973099-158973099 on Build GRCh38
rs748736465 Chr.4: 158973077-158973077 on Build GRCh38
rs970736295 Chr.4: 158973020-158973020 on Build GRCh38
rs190755355 Chr.4: 158973003-158973003 on Build GRCh38
rs757780416 Chr.4: 158973078-158973078 on Build GRCh38
rs959524618 Chr.4: 158973025-158973025 on Build GRCh38
rs1006311317 Chr.4: 158972982-158972982 on Build GRCh38
rs17037858 Chr.4: 158973125-158973125 on Build GRCh38
rs763083990 Chr.4: 158972974-158972974 on Build GRCh38

View Full Product Details