Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33180483-33180710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA2
Entrez Gene ID
140683
Gene Name
BPI fold containing family A member 2
Gene Aliases
C20orf70, PSP, SPLUNC2, bA49G10.1
UniGene
Hs.125960
Gene Chromosome Location
Chr.20: 33161848-33181417 on Build GRCh38
Transcripts
NM_001319164.1 NM_080574.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1713350 c.730C>T p.Q244* Chr.20: 33180540-33180540 on Build GRCh38
6525737 c.749G>A p.*250* Chr.20: 33180559-33180559 on Build GRCh38
6866323 c.710-1G>A p.? Chr.20: 33180519-33180519 on Build GRCh38
4943047 c.735G>T p.L245L Chr.20: 33180545-33180545 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761333694 Chr.20: 33180639-33180644 on Build GRCh38
rs1043394088 Chr.20: 33180520-33180520 on Build GRCh38
rs867940175 Chr.20: 33180615-33180615 on Build GRCh38
rs201682583 Chr.20: 33180571-33180571 on Build GRCh38
rs759733034 Chr.20: 33180537-33180538 on Build GRCh38
rs930263305 Chr.20: 33180658-33180658 on Build GRCh38
rs370478978 Chr.20: 33180590-33180590 on Build GRCh38
rs755614155 Chr.20: 33180551-33180551 on Build GRCh38
rs772570163 Chr.20: 33180564-33180564 on Build GRCh38
rs745343196 Chr.20: 33180550-33180550 on Build GRCh38
rs747462850 Chr.20: 33180567-33180567 on Build GRCh38
rs921685449 Chr.20: 33180653-33180653 on Build GRCh38
rs374708789 Chr.20: 33180588-33180588 on Build GRCh38
rs781651535 Chr.20: 33180667-33180667 on Build GRCh38
rs373405970 Chr.20: 33180565-33180565 on Build GRCh38
rs761710906 Chr.20: 33180603-33180603 on Build GRCh38
rs199534853 Chr.20: 33180537-33180537 on Build GRCh38
rs748593348 Chr.20: 33180560-33180560 on Build GRCh38
rs1023689683 Chr.20: 33180524-33180524 on Build GRCh38
rs756986249 Chr.20: 33180638-33180638 on Build GRCh38
rs774222642 Chr.20: 33180543-33180554 on Build GRCh38
rs761713817 Chr.20: 33180557-33180557 on Build GRCh38
rs771034655 Chr.20: 33180577-33180577 on Build GRCh38
rs377661210 Chr.20: 33180635-33180635 on Build GRCh38
rs768491481 Chr.20: 33180539-33180541 on Build GRCh38
rs751381010 Chr.20: 33180602-33180602 on Build GRCh38
rs571582152 Chr.20: 33180555-33180555 on Build GRCh38
rs776887632 Chr.20: 33180536-33180537 on Build GRCh38
rs776951258 Chr.20: 33180578-33180578 on Build GRCh38
rs143444594 Chr.20: 33180525-33180525 on Build GRCh38
rs1038777267 Chr.20: 33180651-33180651 on Build GRCh38
rs371156549 Chr.20: 33180586-33180586 on Build GRCh38
rs34968847 Chr.20: 33180557-33180557 on Build GRCh38
rs147944008 Chr.20: 33180581-33180581 on Build GRCh38

View Full Product Details