Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119097337-119097582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPAGT1
Entrez Gene ID
1798
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
Gene Aliases
ALG7, CDG-Ij, CDG1J, CMS13, CMSTA2, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT
UniGene
Hs.524081
Gene Chromosome Location
Chr.11: 119096503-119102414 on Build GRCh38
Transcripts
NM_001382.3 XM_005271422.2 XM_011542648.1 XM_017017293.1 XM_017017294.1 XM_017017295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6648767 c.918-9C>T p.? Chr.11: 119097560-119097560 on Build GRCh38
7080320 c.663C>T p.F221F Chr.11: 119097485-119097485 on Build GRCh38
7119857 c.930G>A p.K310K Chr.11: 119097539-119097539 on Build GRCh38
6648768 c.597-9C>T p.? Chr.11: 119097560-119097560 on Build GRCh38
7080319 c.984C>T p.F328F Chr.11: 119097485-119097485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003269168 Chr.11: 119097530-119097530 on Build GRCh38
rs147104217 Chr.11: 119097394-119097394 on Build GRCh38
rs754036071 Chr.11: 119097488-119097489 on Build GRCh38
rs763140128 Chr.11: 119097415-119097415 on Build GRCh38
rs941907857 Chr.11: 119097477-119097477 on Build GRCh38
rs780497256 Chr.11: 119097480-119097480 on Build GRCh38
rs371389351 Chr.11: 119097544-119097544 on Build GRCh38
rs141849365 Chr.11: 119097374-119097374 on Build GRCh38
rs1042544648 Chr.11: 119097482-119097482 on Build GRCh38
rs767497123 Chr.11: 119097418-119097418 on Build GRCh38
rs1045143648 Chr.11: 119097440-119097440 on Build GRCh38
rs201656540 Chr.11: 119097555-119097555 on Build GRCh38
rs756851791 Chr.11: 119097453-119097453 on Build GRCh38
rs545736353 Chr.11: 119097526-119097526 on Build GRCh38
rs764995440 Chr.11: 119097474-119097474 on Build GRCh38
rs757905259 Chr.11: 119097479-119097479 on Build GRCh38
rs760501264 Chr.11: 119097437-119097437 on Build GRCh38
rs775602137 Chr.11: 119097427-119097427 on Build GRCh38
rs531346717 Chr.11: 119097559-119097559 on Build GRCh38
rs748605109 Chr.11: 119097523-119097523 on Build GRCh38
rs747534998 Chr.11: 119097491-119097491 on Build GRCh38
rs781380261 Chr.11: 119097498-119097498 on Build GRCh38
rs138544311 Chr.11: 119097475-119097475 on Build GRCh38
rs191573465 Chr.11: 119097556-119097556 on Build GRCh38
rs763778104 Chr.11: 119097438-119097438 on Build GRCh38
rs755527720 Chr.11: 119097493-119097493 on Build GRCh38
rs753712106 Chr.11: 119097444-119097444 on Build GRCh38
rs550478594 Chr.11: 119097497-119097497 on Build GRCh38

View Full Product Details