Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136327531-136327774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPSM1
Entrez Gene ID
26086
Gene Name
G-protein signaling modulator 1
Gene Aliases
AGS3
UniGene
Hs.239370
Gene Chromosome Location
Chr.9: 136327476-136359605 on Build GRCh38
Transcripts
NM_001145638.2 NM_015597.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5960068 c.14C>A p.A5D Chr.9: 136327709-136327709 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs575324588 Chr.9: 136327558-136327558 on Build GRCh38
rs781914806 Chr.9: 136327607-136327607 on Build GRCh38
rs528405359 Chr.9: 136327640-136327640 on Build GRCh38
rs28495820 Chr.9: 136327718-136327718 on Build GRCh38
rs561078522 Chr.9: 136327587-136327587 on Build GRCh38
rs543015996 Chr.9: 136327562-136327562 on Build GRCh38
rs782059485 Chr.9: 136327713-136327713 on Build GRCh38

View Full Product Details