Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48963035-48963274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSHR
Entrez Gene ID
2492
Gene Name
follicle stimulating hormone receptor
Gene Aliases
FSHR1, FSHRO, LGR1, ODG1
UniGene
Hs.1428
Gene Chromosome Location
Chr.2: 48953161-49154527 on Build GRCh38
Transcripts
NM_000145.3 NM_181446.2 XM_011532733.2 XM_011532734.2 XM_011532735.2 XM_011532736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5686091 c.1665A>T p.T555T Chr.2: 48963156-48963156 on Build GRCh38
394499 c.1618C>A p.L540M Chr.2: 48963203-48963203 on Build GRCh38
721656 c.1639G>T p.G547C Chr.2: 48963182-48963182 on Build GRCh38
3972266 c.1653C>A p.H551Q Chr.2: 48963168-48963168 on Build GRCh38
4930752 c.1695T>C p.S565S Chr.2: 48963126-48963126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757909841 Chr.2: 48963225-48963225 on Build GRCh38
rs991177976 Chr.2: 48963207-48963207 on Build GRCh38
rs1020301126 Chr.2: 48963145-48963145 on Build GRCh38
rs368367400 Chr.2: 48963178-48963178 on Build GRCh38
rs145815060 Chr.2: 48963217-48963217 on Build GRCh38
rs746810761 Chr.2: 48963093-48963093 on Build GRCh38
rs200144377 Chr.2: 48963157-48963157 on Build GRCh38
rs374191266 Chr.2: 48963152-48963152 on Build GRCh38
rs750176361 Chr.2: 48963088-48963088 on Build GRCh38
rs386833511 Chr.2: 48963097-48963097 on Build GRCh38
rs766158543 Chr.2: 48963162-48963162 on Build GRCh38
rs776525084 Chr.2: 48963163-48963163 on Build GRCh38
rs267599403 Chr.2: 48963241-48963241 on Build GRCh38
rs769427769 Chr.2: 48963074-48963074 on Build GRCh38
rs764638641 Chr.2: 48963215-48963215 on Build GRCh38
rs748888573 Chr.2: 48963142-48963142 on Build GRCh38
rs759651692 Chr.2: 48963171-48963171 on Build GRCh38
rs752868051 Chr.2: 48963115-48963115 on Build GRCh38
rs200482566 Chr.2: 48963137-48963137 on Build GRCh38
rs775127668 Chr.2: 48963110-48963110 on Build GRCh38
rs768038206 Chr.2: 48963069-48963069 on Build GRCh38
rs761625807 Chr.2: 48963078-48963078 on Build GRCh38
rs781669876 Chr.2: 48963234-48963234 on Build GRCh38
rs770903417 Chr.2: 48963253-48963253 on Build GRCh38
rs866519552 Chr.2: 48963072-48963072 on Build GRCh38
rs754540694 Chr.2: 48963238-48963238 on Build GRCh38
rs201840639 Chr.2: 48963081-48963081 on Build GRCh38
rs113877695 Chr.2: 48963091-48963091 on Build GRCh38
rs138281715 Chr.2: 48963245-48963245 on Build GRCh38
rs752677466 Chr.2: 48963182-48963182 on Build GRCh38
rs781730288 Chr.2: 48963141-48963141 on Build GRCh38
rs201078200 Chr.2: 48963226-48963226 on Build GRCh38
rs757713421 Chr.2: 48963138-48963138 on Build GRCh38
rs28928871 Chr.2: 48963122-48963122 on Build GRCh38
rs386833512 Chr.2: 48963061-48963061 on Build GRCh38
rs773145656 Chr.2: 48963153-48963153 on Build GRCh38
rs371488439 Chr.2: 48963181-48963181 on Build GRCh38
rs758780668 Chr.2: 48963126-48963126 on Build GRCh38
rs778629351 Chr.2: 48963239-48963239 on Build GRCh38
rs375475417 Chr.2: 48963103-48963103 on Build GRCh38
rs151109524 Chr.2: 48963095-48963095 on Build GRCh38
rs1050610586 Chr.2: 48963123-48963123 on Build GRCh38
rs121909660 Chr.2: 48963104-48963104 on Build GRCh38
rs951624168 Chr.2: 48963161-48963161 on Build GRCh38
rs6167 Chr.2: 48963249-48963249 on Build GRCh38
rs996873705 Chr.2: 48963117-48963117 on Build GRCh38
rs746295796 Chr.2: 48963235-48963235 on Build GRCh38
rs746917603 Chr.2: 48963252-48963252 on Build GRCh38
rs150235567 Chr.2: 48963151-48963151 on Build GRCh38
rs750536074 Chr.2: 48963113-48963114 on Build GRCh38
rs773713354 Chr.2: 48963149-48963149 on Build GRCh38
rs121909664 Chr.2: 48963187-48963187 on Build GRCh38
rs201903308 Chr.2: 48963144-48963144 on Build GRCh38
rs778457986 Chr.2: 48963133-48963133 on Build GRCh38
rs778824635 Chr.2: 48963111-48963111 on Build GRCh38

View Full Product Details