Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40881528-40881785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2A7
Entrez Gene ID
1549
Gene Name
cytochrome P450 family 2 subfamily A member 7
Gene Aliases
CPA7, CPAD, CYP2A, CYPIIA7, P450-IIA4
UniGene
Hs.719890
Gene Chromosome Location
Chr.19: 40875439-40885952 on Build GRCh38
Transcripts
NM_000764.2 NM_030589.2 XM_017026383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1721779 c.318C>T p.T106T Chr.19: 40881614-40881614 on Build GRCh38
297941 c.312A>C p.Q104H Chr.19: 40881620-40881620 on Build GRCh38
6201565 c.317C>T p.T106I Chr.19: 40881615-40881615 on Build GRCh38
7338123 c.268G>A p.A90T Chr.19: 40881664-40881664 on Build GRCh38
4591719 c.190T>C p.C64R Chr.19: 40881742-40881742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772593210 Chr.19: 40881558-40881558 on Build GRCh38
rs558900936 Chr.19: 40881697-40881697 on Build GRCh38
rs550633601 Chr.19: 40881676-40881676 on Build GRCh38
rs375415174 Chr.19: 40881730-40881730 on Build GRCh38
rs750626429 Chr.19: 40881611-40881611 on Build GRCh38
rs745378046 Chr.19: 40881732-40881732 on Build GRCh38
rs112051160 Chr.19: 40881635-40881635 on Build GRCh38
rs756807315 Chr.19: 40881691-40881691 on Build GRCh38
rs745583378 Chr.19: 40881695-40881695 on Build GRCh38
rs764297504 Chr.19: 40881594-40881594 on Build GRCh38
rs776761737 Chr.19: 40881701-40881701 on Build GRCh38
rs773217412 Chr.19: 40881743-40881743 on Build GRCh38
rs746242104 Chr.19: 40881547-40881547 on Build GRCh38
rs753984557 Chr.19: 40881598-40881598 on Build GRCh38
rs369565698 Chr.19: 40881731-40881731 on Build GRCh38
rs530716589 Chr.19: 40881610-40881610 on Build GRCh38
rs200907719 Chr.19: 40881631-40881631 on Build GRCh38
rs367702429 Chr.19: 40881637-40881637 on Build GRCh38
rs770992387 Chr.19: 40881746-40881746 on Build GRCh38
rs762973238 Chr.19: 40881744-40881744 on Build GRCh38
rs747618272 Chr.19: 40881626-40881626 on Build GRCh38
rs146716059 Chr.19: 40881726-40881726 on Build GRCh38
rs760465948 Chr.19: 40881652-40881652 on Build GRCh38
rs187665825 Chr.19: 40881553-40881553 on Build GRCh38
rs534988246 Chr.19: 40881703-40881703 on Build GRCh38
rs545708572 Chr.19: 40881620-40881620 on Build GRCh38
rs760968318 Chr.19: 40881760-40881760 on Build GRCh38
rs201180533 Chr.19: 40881570-40881570 on Build GRCh38
rs748155186 Chr.19: 40881741-40881741 on Build GRCh38
rs768613254 Chr.19: 40881735-40881735 on Build GRCh38
rs779598490 Chr.19: 40881734-40881734 on Build GRCh38
rs746815604 Chr.19: 40881698-40881698 on Build GRCh38
rs763031354 Chr.19: 40881705-40881705 on Build GRCh38
rs906836044 Chr.19: 40881710-40881710 on Build GRCh38
rs751239298 Chr.19: 40881766-40881766 on Build GRCh38
rs750716240 Chr.19: 40881672-40881672 on Build GRCh38
rs372343732 Chr.19: 40881604-40881604 on Build GRCh38
rs376196833 Chr.19: 40881568-40881568 on Build GRCh38
rs142733902 Chr.19: 40881673-40881673 on Build GRCh38
rs754879714 Chr.19: 40881575-40881575 on Build GRCh38
rs1002533572 Chr.19: 40881714-40881714 on Build GRCh38
rs370301485 Chr.19: 40881549-40881549 on Build GRCh38
rs200248426 Chr.19: 40881702-40881702 on Build GRCh38
rs770386243 Chr.19: 40881634-40881634 on Build GRCh38
rs778360387 Chr.19: 40881694-40881694 on Build GRCh38
rs781686376 Chr.19: 40881687-40881687 on Build GRCh38
rs776564344 Chr.19: 40881736-40881736 on Build GRCh38
rs774484257 Chr.19: 40881750-40881750 on Build GRCh38
rs10425169 Chr.19: 40881742-40881742 on Build GRCh38
rs999786638 Chr.19: 40881614-40881614 on Build GRCh38
rs753044172 Chr.19: 40881755-40881755 on Build GRCh38
rs748799988 Chr.19: 40881633-40881633 on Build GRCh38
rs765388487 Chr.19: 40881667-40881667 on Build GRCh38
rs10425185 Chr.19: 40881764-40881764 on Build GRCh38
rs374512565 Chr.19: 40881576-40881576 on Build GRCh38
rs771938642 Chr.19: 40881641-40881641 on Build GRCh38
rs762008461 Chr.19: 40881666-40881666 on Build GRCh38
rs10425176 Chr.19: 40881751-40881751 on Build GRCh38
rs769386794 Chr.19: 40881556-40881556 on Build GRCh38
rs1034196943 Chr.19: 40881557-40881557 on Build GRCh38
rs770582541 Chr.19: 40881567-40881567 on Build GRCh38
rs371905883 Chr.19: 40881657-40881657 on Build GRCh38
rs1019966855 Chr.19: 40881709-40881709 on Build GRCh38
rs537620574 Chr.19: 40881686-40881686 on Build GRCh38
rs763946241 Chr.19: 40881654-40881654 on Build GRCh38
rs757897810 Chr.19: 40881763-40881763 on Build GRCh38
rs764621516 Chr.19: 40881762-40881762 on Build GRCh38
rs202195291 Chr.19: 40881674-40881674 on Build GRCh38
rs568885914 Chr.19: 40881680-40881680 on Build GRCh38
rs759355788 Chr.19: 40881569-40881569 on Build GRCh38
rs570768596 Chr.19: 40881699-40881699 on Build GRCh38
rs748603752 Chr.19: 40881689-40881689 on Build GRCh38
rs757888377 Chr.19: 40881733-40881733 on Build GRCh38
rs775139152 Chr.19: 40881646-40881646 on Build GRCh38
rs374034412 Chr.19: 40881552-40881552 on Build GRCh38
rs191930445 Chr.19: 40881630-40881630 on Build GRCh38
rs780251177 Chr.19: 40881622-40881622 on Build GRCh38
rs766513839 Chr.19: 40881706-40881706 on Build GRCh38
rs10425150 Chr.19: 40881715-40881715 on Build GRCh38
rs368931842 Chr.19: 40881577-40881577 on Build GRCh38
rs376940360 Chr.19: 40881682-40881682 on Build GRCh38
rs981151261 Chr.19: 40881554-40881554 on Build GRCh38

View Full Product Details