Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 34942494-34942763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L1
Entrez Gene ID
23333
Gene Name
dpy-19 like 1 (C. elegans)
Gene Aliases
-
UniGene
Hs.408623
Gene Chromosome Location
Chr.7: 34928881-35038041 on Build GRCh38
Transcripts
NM_015283.1 XM_011515245.2 XM_011515246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3638371 c.1342C>T p.H448Y Chr.7: 34942623-34942623 on Build GRCh38
6649061 c.1326-10delT p.? Chr.7: 34942649-34942649 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755987138 Chr.7: 34942687-34942689 on Build GRCh38
rs945552239 Chr.7: 34942550-34942554 on Build GRCh38
rs778993164 Chr.7: 34942591-34942591 on Build GRCh38
rs779642434 Chr.7: 34942687-34942688 on Build GRCh38
rs201857929 Chr.7: 34942524-34942524 on Build GRCh38
rs530703965 Chr.7: 34942556-34942556 on Build GRCh38
rs749519299 Chr.7: 34942583-34942583 on Build GRCh38
rs770684786 Chr.7: 34942608-34942608 on Build GRCh38
rs759168043 Chr.7: 34942633-34942633 on Build GRCh38
rs750767487 Chr.7: 34942570-34942570 on Build GRCh38
rs754168784 Chr.7: 34942688-34942688 on Build GRCh38
rs914059939 Chr.7: 34942528-34942528 on Build GRCh38
rs767296117 Chr.7: 34942610-34942611 on Build GRCh38
rs780604216 Chr.7: 34942582-34942582 on Build GRCh38
rs750927003 Chr.7: 34942664-34942664 on Build GRCh38
rs576853526 Chr.7: 34942694-34942694 on Build GRCh38
rs567104360 Chr.7: 34942723-34942723 on Build GRCh38
rs747000764 Chr.7: 34942607-34942607 on Build GRCh38
rs756522492 Chr.7: 34942575-34942575 on Build GRCh38
rs989578460 Chr.7: 34942522-34942522 on Build GRCh38
rs755161945 Chr.7: 34942689-34942689 on Build GRCh38
rs371195193 Chr.7: 34942661-34942661 on Build GRCh38
rs748183004 Chr.7: 34942595-34942595 on Build GRCh38
rs1030174865 Chr.7: 34942711-34942711 on Build GRCh38
rs377706763 Chr.7: 34942660-34942660 on Build GRCh38
rs766497369 Chr.7: 34942719-34942719 on Build GRCh38
rs745348198 Chr.7: 34942529-34942529 on Build GRCh38
rs541310786 Chr.7: 34942695-34942695 on Build GRCh38
rs776465135 Chr.7: 34942609-34942609 on Build GRCh38
rs756751097 Chr.7: 34942667-34942667 on Build GRCh38
rs986085149 Chr.7: 34942734-34942734 on Build GRCh38
rs376278279 Chr.7: 34942690-34942690 on Build GRCh38
rs764798168 Chr.7: 34942643-34942643 on Build GRCh38
rs766838815 Chr.7: 34942677-34942677 on Build GRCh38
rs202077354 Chr.7: 34942599-34942599 on Build GRCh38
rs775021965 Chr.7: 34942647-34942647 on Build GRCh38
rs750401102 Chr.7: 34942649-34942649 on Build GRCh38
rs755215893 Chr.7: 34942585-34942585 on Build GRCh38

View Full Product Details