Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 564476-564723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR35
Entrez Gene ID
146325
Gene Name
proline rich 35
Gene Aliases
C16orf11, LA16c-366D1.2
UniGene
Hs.632220
Gene Chromosome Location
Chr.16: 559786-565529 on Build GRCh38
Transcripts
NM_145270.2 XM_017022959.1 XM_017022960.1 XM_017022961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358095 c.1112G>C p.W371S Chr.16: 564703-564703 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs963576795 Chr.16: 564581-564581 on Build GRCh38
rs12919048 Chr.16: 564630-564630 on Build GRCh38
rs571927525 Chr.16: 564527-564527 on Build GRCh38
rs557447735 Chr.16: 564528-564528 on Build GRCh38
rs375903431 Chr.16: 564602-564605 on Build GRCh38
rs530884571 Chr.16: 564646-564646 on Build GRCh38
rs970174567 Chr.16: 564607-564607 on Build GRCh38
rs796526444 Chr.16: 564605-564605 on Build GRCh38
rs541737773 Chr.16: 564557-564557 on Build GRCh38
rs777776333 Chr.16: 564660-564660 on Build GRCh38
rs111764990 Chr.16: 564601-564604 on Build GRCh38
rs900249549 Chr.16: 564537-564537 on Build GRCh38
rs575042468 Chr.16: 564623-564623 on Build GRCh38
rs952029529 Chr.16: 564674-564674 on Build GRCh38
rs572911571 Chr.16: 564543-564543 on Build GRCh38
rs1018739101 Chr.16: 564641-564641 on Build GRCh38
rs1023849080 Chr.16: 564588-564588 on Build GRCh38
rs914823607 Chr.16: 564657-564657 on Build GRCh38
rs895528202 Chr.16: 564553-564553 on Build GRCh38
rs746785902 Chr.16: 564684-564684 on Build GRCh38
rs990665229 Chr.16: 564587-564587 on Build GRCh38
rs893333630 Chr.16: 564525-564525 on Build GRCh38
rs199532631 Chr.16: 564703-564703 on Build GRCh38
rs758193893 Chr.16: 564549-564549 on Build GRCh38
rs886080720 Chr.16: 564550-564550 on Build GRCh38
rs940866967 Chr.16: 564544-564544 on Build GRCh38
rs900636038 Chr.16: 564678-564678 on Build GRCh38
rs949799821 Chr.16: 564556-564556 on Build GRCh38
rs754466571 Chr.16: 564629-564629 on Build GRCh38
rs560138694 Chr.16: 564565-564565 on Build GRCh38
rs908111964 Chr.16: 564676-564676 on Build GRCh38
rs933101331 Chr.16: 564699-564699 on Build GRCh38
rs1039207936 Chr.16: 564666-564666 on Build GRCh38
rs989091387 Chr.16: 564645-564645 on Build GRCh38
rs997637373 Chr.16: 564540-564540 on Build GRCh38
rs142577477 Chr.16: 564624-564624 on Build GRCh38
rs538926505 Chr.16: 564539-564539 on Build GRCh38
rs985235795 Chr.16: 564675-564675 on Build GRCh38
rs996231588 Chr.16: 564652-564652 on Build GRCh38
rs60243750 Chr.16: 564658-564658 on Build GRCh38

View Full Product Details