Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160999751-160999982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F11R
Entrez Gene ID
50848
Gene Name
F11 receptor
Gene Aliases
CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1
UniGene
Hs.517293
Gene Chromosome Location
Chr.1: 160995211-161021343 on Build GRCh38
Transcripts
NM_016946.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6657665 c.671C>T p.T224I Chr.1: 160999911-160999911 on Build GRCh38
4901560 c.622G>A p.G208R Chr.1: 160999948-160999948 on Build GRCh38
4401713 c.593T>C p.V198A Chr.1: 160999977-160999977 on Build GRCh38
4502061 c.603C>T p.P201P Chr.1: 160999967-160999967 on Build GRCh38
5165466 c.674C>A p.P225H Chr.1: 160999908-160999908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370646737 Chr.1: 160999900-160999900 on Build GRCh38
rs368748813 Chr.1: 160999927-160999927 on Build GRCh38
rs772481019 Chr.1: 160999957-160999957 on Build GRCh38
rs750230279 Chr.1: 160999895-160999895 on Build GRCh38
rs985553315 Chr.1: 160999886-160999886 on Build GRCh38
rs201801022 Chr.1: 160999792-160999792 on Build GRCh38
rs746425664 Chr.1: 160999840-160999840 on Build GRCh38
rs746183098 Chr.1: 160999945-160999945 on Build GRCh38
rs751059901 Chr.1: 160999908-160999908 on Build GRCh38
rs183326320 Chr.1: 160999851-160999851 on Build GRCh38
rs34601197 Chr.1: 160999835-160999836 on Build GRCh38
rs894859437 Chr.1: 160999801-160999801 on Build GRCh38
rs755819660 Chr.1: 160999926-160999926 on Build GRCh38
rs371284289 Chr.1: 160999809-160999809 on Build GRCh38
rs373885875 Chr.1: 160999842-160999842 on Build GRCh38
rs774839104 Chr.1: 160999837-160999837 on Build GRCh38
rs758274072 Chr.1: 160999899-160999899 on Build GRCh38
rs790056 Chr.1: 160999795-160999795 on Build GRCh38
rs775448381 Chr.1: 160999850-160999850 on Build GRCh38
rs199703485 Chr.1: 160999888-160999888 on Build GRCh38
rs140170242 Chr.1: 160999821-160999821 on Build GRCh38
rs142730492 Chr.1: 160999776-160999776 on Build GRCh38
rs780949710 Chr.1: 160999793-160999793 on Build GRCh38
rs978177241 Chr.1: 160999835-160999835 on Build GRCh38
rs752452076 Chr.1: 160999916-160999916 on Build GRCh38
rs553706719 Chr.1: 160999904-160999904 on Build GRCh38
rs533878408 Chr.1: 160999893-160999893 on Build GRCh38
rs147266047 Chr.1: 160999887-160999887 on Build GRCh38
rs764335444 Chr.1: 160999885-160999885 on Build GRCh38
rs149862476 Chr.1: 160999913-160999913 on Build GRCh38
rs1010472713 Chr.1: 160999921-160999921 on Build GRCh38
rs780826713 Chr.1: 160999915-160999915 on Build GRCh38
rs746469278 Chr.1: 160999788-160999788 on Build GRCh38
rs146072377 Chr.1: 160999785-160999785 on Build GRCh38
rs201003339 Chr.1: 160999832-160999832 on Build GRCh38
rs374519076 Chr.1: 160999775-160999775 on Build GRCh38
rs779498236 Chr.1: 160999787-160999787 on Build GRCh38
rs764986576 Chr.1: 160999777-160999777 on Build GRCh38

View Full Product Details