Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49016446-49016677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DALRD3
Entrez Gene ID
55152
Gene Name
DALR anticodon binding domain containing 3
Gene Aliases
-
UniGene
Hs.654815
Gene Chromosome Location
Chr.3: 49015399-49021386 on Build GRCh38
Transcripts
NM_001009996.2 NM_001276405.1 NM_018114.5 XM_005265269.3 XM_006713219.2 XM_011533891.1 XM_011533892.1 XM_011533893.1 XM_017006723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991582 c.1024A>G p.K342E Chr.3: 49016651-49016651 on Build GRCh38
4118375 c.563-1G>A p.? Chr.3: 49016512-49016512 on Build GRCh38
1495550 c.533C>T p.A178V Chr.3: 49016641-49016641 on Build GRCh38
7116653 c.563-1G>C p.? Chr.3: 49016512-49016512 on Build GRCh38
4635012 c.566C>T p.P189L Chr.3: 49016508-49016508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759163780 Chr.3: 49016582-49016582 on Build GRCh38
rs755249153 Chr.3: 49016626-49016626 on Build GRCh38
rs757089849 Chr.3: 49016569-49016571 on Build GRCh38
rs142612108 Chr.3: 49016551-49016551 on Build GRCh38
rs774789258 Chr.3: 49016531-49016531 on Build GRCh38
rs780260546 Chr.3: 49016615-49016615 on Build GRCh38
rs973007158 Chr.3: 49016589-49016589 on Build GRCh38
rs778924961 Chr.3: 49016480-49016480 on Build GRCh38
rs202087735 Chr.3: 49016609-49016609 on Build GRCh38
rs1017029392 Chr.3: 49016537-49016537 on Build GRCh38
rs762479241 Chr.3: 49016567-49016567 on Build GRCh38
rs770474292 Chr.3: 49016658-49016658 on Build GRCh38
rs781123128 Chr.3: 49016521-49016521 on Build GRCh38
rs777722603 Chr.3: 49016634-49016634 on Build GRCh38
rs759250055 Chr.3: 49016660-49016660 on Build GRCh38
rs754196252 Chr.3: 49016584-49016584 on Build GRCh38
rs770517582 Chr.3: 49016572-49016572 on Build GRCh38
rs747922020 Chr.3: 49016630-49016630 on Build GRCh38
rs762228824 Chr.3: 49016590-49016590 on Build GRCh38
rs765717665 Chr.3: 49016608-49016608 on Build GRCh38
rs372925729 Chr.3: 49016632-49016632 on Build GRCh38
rs560679760 Chr.3: 49016489-49016489 on Build GRCh38
rs777839460 Chr.3: 49016537-49016538 on Build GRCh38
rs766982007 Chr.3: 49016504-49016504 on Build GRCh38
rs145394428 Chr.3: 49016485-49016485 on Build GRCh38
rs768623169 Chr.3: 49016505-49016505 on Build GRCh38
rs755752013 Chr.3: 49016477-49016480 on Build GRCh38
rs559963267 Chr.3: 49016503-49016503 on Build GRCh38
rs370665532 Chr.3: 49016547-49016547 on Build GRCh38
rs199627545 Chr.3: 49016575-49016575 on Build GRCh38
rs758546850 Chr.3: 49016613-49016613 on Build GRCh38
rs561169716 Chr.3: 49016583-49016583 on Build GRCh38
rs749285887 Chr.3: 49016562-49016563 on Build GRCh38
rs367655503 Chr.3: 49016624-49016624 on Build GRCh38
rs781141869 Chr.3: 49016627-49016627 on Build GRCh38
rs778883348 Chr.3: 49016577-49016577 on Build GRCh38
rs575526933 Chr.3: 49016599-49016599 on Build GRCh38
rs749144961 Chr.3: 49016655-49016655 on Build GRCh38
rs772615625 Chr.3: 49016552-49016552 on Build GRCh38

View Full Product Details