Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86899913-86900186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR1A
Entrez Gene ID
657
Gene Name
bone morphogenetic protein receptor type 1A
Gene Aliases
10q23del, ACVRLK3, ALK3, CD292, SKR5
UniGene
Hs.524477
Gene Chromosome Location
Chr.10: 86755786-86927969 on Build GRCh38
Transcripts
NM_004329.2 XM_011540103.2 XM_011540104.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612023 c.514A>G p.S172G Chr.10: 86900110-86900110 on Build GRCh38
6983738 c.441T>A p.F147L Chr.10: 86900037-86900037 on Build GRCh38
6939124 c.442G>A p.D148N Chr.10: 86900038-86900038 on Build GRCh38
1967779 c.455G>C p.R152P Chr.10: 86900051-86900051 on Build GRCh38
6960090 c.436delT p.F147fs*18 Chr.10: 86900032-86900032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377733546 Chr.10: 86900107-86900107 on Build GRCh38
rs876659613 Chr.10: 86900024-86900024 on Build GRCh38
rs1057518941 Chr.10: 86900098-86900100 on Build GRCh38
rs767249542 Chr.10: 86900153-86900153 on Build GRCh38
rs876658490 Chr.10: 86900130-86900130 on Build GRCh38
rs367550999 Chr.10: 86900139-86900139 on Build GRCh38
rs778615896 Chr.10: 86900100-86900100 on Build GRCh38
rs148331454 Chr.10: 86899997-86899997 on Build GRCh38
rs749736191 Chr.10: 86900148-86900148 on Build GRCh38
rs372499777 Chr.10: 86899975-86899975 on Build GRCh38
rs567009904 Chr.10: 86900051-86900051 on Build GRCh38
rs759223499 Chr.10: 86900070-86900070 on Build GRCh38
rs370091063 Chr.10: 86900120-86900120 on Build GRCh38
rs587778112 Chr.10: 86900033-86900033 on Build GRCh38
rs762900005 Chr.10: 86900032-86900032 on Build GRCh38
rs964724409 Chr.10: 86900134-86900134 on Build GRCh38
rs371243894 Chr.10: 86900036-86900036 on Build GRCh38
rs763210375 Chr.10: 86900046-86900046 on Build GRCh38
rs372259392 Chr.10: 86900150-86900150 on Build GRCh38
rs781286980 Chr.10: 86900108-86900108 on Build GRCh38
rs142965096 Chr.10: 86900086-86900086 on Build GRCh38
rs879174600 Chr.10: 86900028-86900028 on Build GRCh38
rs863224721 Chr.10: 86900115-86900115 on Build GRCh38
rs200951235 Chr.10: 86900095-86900095 on Build GRCh38
rs763554505 Chr.10: 86900102-86900103 on Build GRCh38
rs763934014 Chr.10: 86900091-86900091 on Build GRCh38
rs751260747 Chr.10: 86900066-86900066 on Build GRCh38
rs567576725 Chr.10: 86900149-86900149 on Build GRCh38
rs7920259 Chr.10: 86900001-86900001 on Build GRCh38
rs552061352 Chr.10: 86899974-86899974 on Build GRCh38
rs780127152 Chr.10: 86899991-86899991 on Build GRCh38
rs555873943 Chr.10: 86900105-86900105 on Build GRCh38
rs587782334 Chr.10: 86900022-86900022 on Build GRCh38
rs145101532 Chr.10: 86900074-86900074 on Build GRCh38
rs11818239 Chr.10: 86900031-86900031 on Build GRCh38
rs747266339 Chr.10: 86900106-86900106 on Build GRCh38
rs976070262 Chr.10: 86900138-86900138 on Build GRCh38
rs773689557 Chr.10: 86900042-86900042 on Build GRCh38
rs775347111 Chr.10: 86899983-86899984 on Build GRCh38
rs971636078 Chr.10: 86900092-86900092 on Build GRCh38
rs878854669 Chr.10: 86900105-86900107 on Build GRCh38
rs772406542 Chr.10: 86899984-86899984 on Build GRCh38
rs767157505 Chr.10: 86900071-86900071 on Build GRCh38
rs878854668 Chr.10: 86900101-86900101 on Build GRCh38
rs770387084 Chr.10: 86900124-86900124 on Build GRCh38

View Full Product Details