Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9732830-9733097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLECL1
Entrez Gene ID
160365
Gene Name
C-type lectin like 1
Gene Aliases
DCAL-1, DCAL1
UniGene
Hs.560087
Gene Chromosome Location
Chr.12: 9715860-9733435 on Build GRCh38
Transcripts
NM_001253750.1 NM_001267701.1 NM_172004.3 XM_011520574.2 XM_017018885.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3466757 c.213C>T p.F71F Chr.12: 9733052-9733052 on Build GRCh38
3688508 c.283C>T p.P95S Chr.12: 9732982-9732982 on Build GRCh38
3664880 c.316+5G>T p.? Chr.12: 9732944-9732944 on Build GRCh38
374091 c.256G>T p.A86S Chr.12: 9733009-9733009 on Build GRCh38
4045583 c.243A>T p.G81G Chr.12: 9733022-9733022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777393740 Chr.12: 9733000-9733000 on Build GRCh38
rs751488466 Chr.12: 9733001-9733001 on Build GRCh38
rs770544921 Chr.12: 9732971-9732971 on Build GRCh38
rs1023499997 Chr.12: 9732969-9732969 on Build GRCh38
rs779195515 Chr.12: 9733037-9733037 on Build GRCh38
rs764110641 Chr.12: 9732920-9732920 on Build GRCh38
rs201053431 Chr.12: 9733063-9733063 on Build GRCh38
rs765775064 Chr.12: 9732872-9732872 on Build GRCh38
rs754111425 Chr.12: 9733029-9733029 on Build GRCh38
rs774280673 Chr.12: 9732897-9732897 on Build GRCh38
rs79809727 Chr.12: 9732982-9732982 on Build GRCh38
rs373212551 Chr.12: 9733046-9733046 on Build GRCh38
rs1035203934 Chr.12: 9732886-9732886 on Build GRCh38
rs748653902 Chr.12: 9733039-9733039 on Build GRCh38
rs762833109 Chr.12: 9732991-9732991 on Build GRCh38
rs757292060 Chr.12: 9732924-9732924 on Build GRCh38
rs779506004 Chr.12: 9732968-9732968 on Build GRCh38
rs752781934 Chr.12: 9732993-9732994 on Build GRCh38
rs750504659 Chr.12: 9733012-9733012 on Build GRCh38
rs772775518 Chr.12: 9733042-9733042 on Build GRCh38
rs1016617605 Chr.12: 9732933-9732933 on Build GRCh38
rs12304510 Chr.12: 9732930-9732930 on Build GRCh38
rs765114525 Chr.12: 9732901-9732901 on Build GRCh38
rs776471903 Chr.12: 9732981-9732981 on Build GRCh38
rs755339740 Chr.12: 9733030-9733030 on Build GRCh38
rs762736859 Chr.12: 9732912-9732912 on Build GRCh38
rs755254210 Chr.12: 9732990-9732990 on Build GRCh38
rs145268017 Chr.12: 9733053-9733053 on Build GRCh38
rs780785881 Chr.12: 9733050-9733050 on Build GRCh38
rs779620162 Chr.12: 9733062-9733062 on Build GRCh38
rs139676111 Chr.12: 9733008-9733008 on Build GRCh38
rs759408171 Chr.12: 9732893-9732893 on Build GRCh38
rs756270202 Chr.12: 9733016-9733016 on Build GRCh38
rs780216106 Chr.12: 9732956-9732956 on Build GRCh38
rs750627618 Chr.12: 9732931-9732931 on Build GRCh38
rs986886282 Chr.12: 9733044-9733044 on Build GRCh38
rs774196156 Chr.12: 9733070-9733070 on Build GRCh38
rs751550011 Chr.12: 9732922-9732922 on Build GRCh38
rs147608426 Chr.12: 9733057-9733057 on Build GRCh38
rs776340468 Chr.12: 9732976-9732976 on Build GRCh38
rs151210429 Chr.12: 9732995-9732995 on Build GRCh38
rs202221764 Chr.12: 9732960-9732960 on Build GRCh38
rs749676733 Chr.12: 9732958-9732958 on Build GRCh38
rs767578087 Chr.12: 9733010-9733010 on Build GRCh38
rs750671734 Chr.12: 9732970-9732970 on Build GRCh38
rs756393100 Chr.12: 9732952-9732952 on Build GRCh38
rs780345412 Chr.12: 9733026-9733026 on Build GRCh38
rs115549012 Chr.12: 9732907-9732907 on Build GRCh38
rs749063416 Chr.12: 9732874-9732874 on Build GRCh38
rs746360704 Chr.12: 9732881-9732881 on Build GRCh38
rs200640673 Chr.12: 9732980-9732980 on Build GRCh38

View Full Product Details