Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13721705-13721966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAR1
Entrez Gene ID
84188
Gene Name
fatty acyl-CoA reductase 1
Gene Aliases
MLSTD2, PFCRD, SDR10E1
UniGene
Hs.501991
Gene Chromosome Location
Chr.11: 13668659-13732346 on Build GRCh38
Transcripts
NM_032228.5 XM_011520400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660544 c.1148G>A p.R383H Chr.11: 13721750-13721750 on Build GRCh38
4019592 c.1147C>T p.R383C Chr.11: 13721749-13721749 on Build GRCh38
3734090 c.1246G>C p.E416Q Chr.11: 13721848-13721848 on Build GRCh38
7367599 c.1253A>T p.K418I Chr.11: 13721855-13721855 on Build GRCh38
6660543 c.1245T>C p.P415P Chr.11: 13721847-13721847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007723601 Chr.11: 13721827-13721827 on Build GRCh38
rs913888324 Chr.11: 13721845-13721845 on Build GRCh38
rs775964151 Chr.11: 13721878-13721880 on Build GRCh38
rs770494479 Chr.11: 13721874-13721875 on Build GRCh38
rs1051997370 Chr.11: 13721810-13721810 on Build GRCh38
rs747108966 Chr.11: 13721743-13721743 on Build GRCh38
rs771913739 Chr.11: 13721892-13721892 on Build GRCh38
rs897509463 Chr.11: 13721764-13721764 on Build GRCh38
rs780116868 Chr.11: 13721735-13721735 on Build GRCh38
rs763501276 Chr.11: 13721883-13721884 on Build GRCh38
rs529821201 Chr.11: 13721882-13721882 on Build GRCh38
rs1043978075 Chr.11: 13721903-13721903 on Build GRCh38
rs140610185 Chr.11: 13721822-13721822 on Build GRCh38
rs746437633 Chr.11: 13721866-13721869 on Build GRCh38
rs774075062 Chr.11: 13721823-13721823 on Build GRCh38
rs768821829 Chr.11: 13721757-13721757 on Build GRCh38
rs34341612 Chr.11: 13721778-13721778 on Build GRCh38
rs945649816 Chr.11: 13721897-13721897 on Build GRCh38
rs764413107 Chr.11: 13721888-13721888 on Build GRCh38
rs781428789 Chr.11: 13721760-13721760 on Build GRCh38
rs904061658 Chr.11: 13721904-13721904 on Build GRCh38

View Full Product Details