Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22629833-22630106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN3
Entrez Gene ID
55909
Gene Name
bridging integrator 3
Gene Aliases
-
UniGene
Hs.433722
Gene Chromosome Location
Chr.8: 22620418-22669148 on Build GRCh38
Transcripts
NM_018688.4 XM_005273573.3 XM_011544586.2 XM_011544587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5686265 c.319G>T p.V107L Chr.8: 22629983-22629983 on Build GRCh38
2786364 c.325G>C p.E109Q Chr.8: 22629977-22629977 on Build GRCh38
421674 c.312G>A p.Q104Q Chr.8: 22629990-22629990 on Build GRCh38
1731773 c.333A>T p.L111F Chr.8: 22629969-22629969 on Build GRCh38
7205671 c.309C>T p.I103I Chr.8: 22629993-22629993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756267807 Chr.8: 22629945-22629945 on Build GRCh38
rs763283093 Chr.8: 22630022-22630023 on Build GRCh38
rs774422889 Chr.8: 22630010-22630010 on Build GRCh38
rs1035417544 Chr.8: 22629935-22629935 on Build GRCh38
rs1049933265 Chr.8: 22630073-22630073 on Build GRCh38
rs781317710 Chr.8: 22630047-22630047 on Build GRCh38
rs374810339 Chr.8: 22630002-22630002 on Build GRCh38
rs754995821 Chr.8: 22629983-22629983 on Build GRCh38
rs374883546 Chr.8: 22630009-22630009 on Build GRCh38
rs577841706 Chr.8: 22629903-22629903 on Build GRCh38
rs765945729 Chr.8: 22630041-22630041 on Build GRCh38
rs770855015 Chr.8: 22630008-22630008 on Build GRCh38
rs980231168 Chr.8: 22629992-22629992 on Build GRCh38
rs968509088 Chr.8: 22629906-22629906 on Build GRCh38
rs749919540 Chr.8: 22629970-22629970 on Build GRCh38
rs369465170 Chr.8: 22630021-22630021 on Build GRCh38
rs772594690 Chr.8: 22630022-22630022 on Build GRCh38
rs750103909 Chr.8: 22630040-22630040 on Build GRCh38
rs372793609 Chr.8: 22630048-22630048 on Build GRCh38
rs773317532 Chr.8: 22629918-22629918 on Build GRCh38
rs915763204 Chr.8: 22629957-22629957 on Build GRCh38
rs533914908 Chr.8: 22629876-22629876 on Build GRCh38
rs751485461 Chr.8: 22629978-22629978 on Build GRCh38
rs764939759 Chr.8: 22630039-22630039 on Build GRCh38
rs6558171 Chr.8: 22629909-22629909 on Build GRCh38
rs982167043 Chr.8: 22629933-22629933 on Build GRCh38
rs754944560 Chr.8: 22630046-22630046 on Build GRCh38
rs781082662 Chr.8: 22629985-22629985 on Build GRCh38
rs753095622 Chr.8: 22629954-22629954 on Build GRCh38
rs761480125 Chr.8: 22629969-22629969 on Build GRCh38
rs376572867 Chr.8: 22630053-22630053 on Build GRCh38
rs6558170 Chr.8: 22629875-22629875 on Build GRCh38
rs776003776 Chr.8: 22629964-22629964 on Build GRCh38
rs376500428 Chr.8: 22630045-22630045 on Build GRCh38
rs747942036 Chr.8: 22629991-22629991 on Build GRCh38
rs201978151 Chr.8: 22629977-22629977 on Build GRCh38
rs770025487 Chr.8: 22629992-22629992 on Build GRCh38
rs371301314 Chr.8: 22629959-22629959 on Build GRCh38
rs901284842 Chr.8: 22629857-22629857 on Build GRCh38
rs777920764 Chr.8: 22629999-22629999 on Build GRCh38
rs769928089 Chr.8: 22629916-22629916 on Build GRCh38
rs757914404 Chr.8: 22629972-22629972 on Build GRCh38
rs575462175 Chr.8: 22629938-22629938 on Build GRCh38
rs940252619 Chr.8: 22630044-22630044 on Build GRCh38
rs539825784 Chr.8: 22630086-22630086 on Build GRCh38
rs932937540 Chr.8: 22630070-22630070 on Build GRCh38
rs904608132 Chr.8: 22630067-22630067 on Build GRCh38
rs767807082 Chr.8: 22629948-22629948 on Build GRCh38
rs760986097 Chr.8: 22630036-22630036 on Build GRCh38
rs755915874 Chr.8: 22630049-22630049 on Build GRCh38
rs553263658 Chr.8: 22629877-22629877 on Build GRCh38
rs774892689 Chr.8: 22629937-22629937 on Build GRCh38
rs775813243 Chr.8: 22630026-22630026 on Build GRCh38
rs560930220 Chr.8: 22630058-22630058 on Build GRCh38
rs749400546 Chr.8: 22630054-22630054 on Build GRCh38
rs367855217 Chr.8: 22629927-22629927 on Build GRCh38
rs572816884 Chr.8: 22630081-22630081 on Build GRCh38

View Full Product Details