Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241804107-241804352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL3ST2
Entrez Gene ID
64090
Gene Name
galactose-3-O-sulfotransferase 2
Gene Aliases
GAL3ST-2, GP3ST
UniGene
Hs.632556
Gene Chromosome Location
Chr.2: 241776825-241804287 on Build GRCh38
Transcripts
NM_022134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4618212 c.1189G>T p.G397W Chr.2: 241804158-241804158 on Build GRCh38
6587441 c.1188delG p.A398fs? Chr.2: 241804157-241804157 on Build GRCh38
5958211 c.1165C>A p.P389T Chr.2: 241804134-241804134 on Build GRCh38
5433146 c.1177A>G p.I393V Chr.2: 241804146-241804146 on Build GRCh38
6401561 c.1156C>A p.P386T Chr.2: 241804125-241804125 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs932966447 Chr.2: 241804333-241804333 on Build GRCh38
rs540275232 Chr.2: 241804275-241804275 on Build GRCh38
rs909053112 Chr.2: 241804282-241804282 on Build GRCh38
rs557660728 Chr.2: 241804191-241804191 on Build GRCh38
rs896242600 Chr.2: 241804308-241804308 on Build GRCh38
rs986556949 Chr.2: 241804248-241804248 on Build GRCh38
rs911112140 Chr.2: 241804261-241804261 on Build GRCh38
rs756212615 Chr.2: 241804255-241804255 on Build GRCh38
rs772071650 Chr.2: 241804179-241804179 on Build GRCh38
rs762129182 Chr.2: 241804329-241804329 on Build GRCh38
rs956775395 Chr.2: 241804235-241804235 on Build GRCh38
rs1046833985 Chr.2: 241804298-241804298 on Build GRCh38
rs759718543 Chr.2: 241804165-241804165 on Build GRCh38
rs546556169 Chr.2: 241804220-241804220 on Build GRCh38
rs1025722734 Chr.2: 241804145-241804145 on Build GRCh38
rs929713824 Chr.2: 241804292-241804292 on Build GRCh38
rs975069929 Chr.2: 241804325-241804325 on Build GRCh38
rs1009727925 Chr.2: 241804190-241804190 on Build GRCh38
rs898368393 Chr.2: 241804274-241804274 on Build GRCh38
rs775257453 Chr.2: 241804185-241804185 on Build GRCh38
rs747351744 Chr.2: 241804127-241804127 on Build GRCh38
rs773700892 Chr.2: 241804168-241804169 on Build GRCh38
rs1038129630 Chr.2: 241804265-241804265 on Build GRCh38
rs528725211 Chr.2: 241804242-241804242 on Build GRCh38
rs112194416 Chr.2: 241804225-241804225 on Build GRCh38
rs963292124 Chr.2: 241804315-241804315 on Build GRCh38
rs993881145 Chr.2: 241804141-241804141 on Build GRCh38
rs933018614 Chr.2: 241804172-241804172 on Build GRCh38
rs776987538 Chr.2: 241804164-241804164 on Build GRCh38
rs1001535119 Chr.2: 241804217-241804218 on Build GRCh38
rs866723965 Chr.2: 241804135-241804135 on Build GRCh38
rs933362810 Chr.2: 241804245-241804245 on Build GRCh38
rs1051509480 Chr.2: 241804174-241804174 on Build GRCh38
rs983286214 Chr.2: 241804281-241804281 on Build GRCh38
rs561637198 Chr.2: 241804307-241804307 on Build GRCh38
rs1043748479 Chr.2: 241804269-241804269 on Build GRCh38
rs536158576 Chr.2: 241804217-241804217 on Build GRCh38
rs767295818 Chr.2: 241804204-241804204 on Build GRCh38
rs744986 Chr.2: 241804236-241804236 on Build GRCh38
rs942630125 Chr.2: 241804262-241804262 on Build GRCh38
rs1013579195 Chr.2: 241804228-241804228 on Build GRCh38
rs373185622 Chr.2: 241804126-241804126 on Build GRCh38
rs376663386 Chr.2: 241804132-241804132 on Build GRCh38
rs1032513148 Chr.2: 241804227-241804227 on Build GRCh38
rs528821591 Chr.2: 241804330-241804330 on Build GRCh38

View Full Product Details