Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 1312817-1313060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXQ1
Entrez Gene ID
94234
Gene Name
forkhead box Q1
Gene Aliases
HFH1
UniGene
Hs.591352
Gene Chromosome Location
Chr.6: 1312440-1314758 on Build GRCh38
Transcripts
NM_033260.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1739283 c.302G>A p.G101D Chr.6: 1313006-1313006 on Build GRCh38
6513712 c.319G>A p.E107K Chr.6: 1313023-1313023 on Build GRCh38
3928084 c.346C>T p.R116W Chr.6: 1313050-1313050 on Build GRCh38
5556214 c.297C>T p.G99G Chr.6: 1313001-1313001 on Build GRCh38
6391427 c.178A>C p.T60P Chr.6: 1312882-1312882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776061715 Chr.6: 1313000-1313000 on Build GRCh38
rs768932766 Chr.6: 1313011-1313011 on Build GRCh38
rs996666315 Chr.6: 1312996-1312996 on Build GRCh38
rs925305500 Chr.6: 1313037-1313037 on Build GRCh38
rs199626291 Chr.6: 1313006-1313006 on Build GRCh38
rs754727576 Chr.6: 1312891-1312891 on Build GRCh38
rs776524301 Chr.6: 1313012-1313012 on Build GRCh38
rs758156468 Chr.6: 1312872-1312872 on Build GRCh38
rs551142495 Chr.6: 1313030-1313030 on Build GRCh38
rs1007934300 Chr.6: 1312927-1312927 on Build GRCh38
rs78460560 Chr.6: 1312843-1312843 on Build GRCh38
rs545254909 Chr.6: 1313015-1313015 on Build GRCh38
rs558903450 Chr.6: 1312994-1312994 on Build GRCh38
rs565575573 Chr.6: 1313017-1313017 on Build GRCh38
rs772548889 Chr.6: 1312999-1312999 on Build GRCh38
rs375808145 Chr.6: 1312984-1312984 on Build GRCh38
rs1041184946 Chr.6: 1312931-1312931 on Build GRCh38
rs936937770 Chr.6: 1312858-1312858 on Build GRCh38
rs200531155 Chr.6: 1313026-1313026 on Build GRCh38
rs780924028 Chr.6: 1313028-1313028 on Build GRCh38
rs762224760 Chr.6: 1312966-1312974 on Build GRCh38
rs751374101 Chr.6: 1313023-1313023 on Build GRCh38
rs765180720 Chr.6: 1312841-1312841 on Build GRCh38
rs9502889 Chr.6: 1312882-1312882 on Build GRCh38
rs781117053 Chr.6: 1312863-1312864 on Build GRCh38
rs746419811 Chr.6: 1312987-1312987 on Build GRCh38
rs9502890 Chr.6: 1312886-1312886 on Build GRCh38
rs762938633 Chr.6: 1313018-1313018 on Build GRCh38
rs573671916 Chr.6: 1312893-1312893 on Build GRCh38
rs561354143 Chr.6: 1313032-1313032 on Build GRCh38
rs771298091 Chr.6: 1312894-1312894 on Build GRCh38
rs370291379 Chr.6: 1313019-1313019 on Build GRCh38
rs765339945 Chr.6: 1313016-1313016 on Build GRCh38
rs765973834 Chr.6: 1312999-1313007 on Build GRCh38

View Full Product Details