Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58153451-58153710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf85
Entrez Gene ID
128602
Gene Name
chromosome 20 open reading frame 85
Gene Aliases
LLC1, bA196N14.1
UniGene
Hs.43977
Gene Chromosome Location
Chr.20: 58150927-58161130 on Build GRCh38
Transcripts
NM_178456.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5584636 c.79C>T p.L27L Chr.20: 58153554-58153554 on Build GRCh38
5006773 c.84G>C p.K28N Chr.20: 58153559-58153559 on Build GRCh38
7358405 c.77G>A p.R26H Chr.20: 58153552-58153552 on Build GRCh38
478331 c.125G>A p.G42E Chr.20: 58153600-58153600 on Build GRCh38
6160416 c.81G>A p.L27L Chr.20: 58153556-58153556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552853226 Chr.20: 58153551-58153551 on Build GRCh38
rs199596476 Chr.20: 58153614-58153614 on Build GRCh38
rs759827043 Chr.20: 58153590-58153590 on Build GRCh38
rs138257522 Chr.20: 58153615-58153615 on Build GRCh38
rs375930485 Chr.20: 58153567-58153567 on Build GRCh38
rs61754559 Chr.20: 58153553-58153553 on Build GRCh38
rs547087983 Chr.20: 58153575-58153575 on Build GRCh38
rs767667785 Chr.20: 58153506-58153506 on Build GRCh38
rs759850244 Chr.20: 58153505-58153505 on Build GRCh38
rs149619510 Chr.20: 58153626-58153626 on Build GRCh38
rs752992462 Chr.20: 58153597-58153597 on Build GRCh38
rs761926698 Chr.20: 58153612-58153612 on Build GRCh38
rs531085959 Chr.20: 58153509-58153509 on Build GRCh38
rs941274577 Chr.20: 58153624-58153624 on Build GRCh38
rs542095898 Chr.20: 58153493-58153493 on Build GRCh38
rs45576936 Chr.20: 58153576-58153576 on Build GRCh38
rs774855883 Chr.20: 58153654-58153654 on Build GRCh38
rs772454910 Chr.20: 58153660-58153660 on Build GRCh38
rs754021691 Chr.20: 58153602-58153602 on Build GRCh38
rs563989715 Chr.20: 58153501-58153501 on Build GRCh38
rs746447084 Chr.20: 58153659-58153659 on Build GRCh38
rs757363332 Chr.20: 58153607-58153607 on Build GRCh38
rs753964252 Chr.20: 58153514-58153514 on Build GRCh38
rs752900872 Chr.20: 58153508-58153508 on Build GRCh38
rs200408259 Chr.20: 58153502-58153502 on Build GRCh38
rs191493508 Chr.20: 58153653-58153653 on Build GRCh38
rs536022748 Chr.20: 58153601-58153601 on Build GRCh38
rs772746059 Chr.20: 58153572-58153572 on Build GRCh38
rs777352750 Chr.20: 58153646-58153646 on Build GRCh38
rs747862895 Chr.20: 58153639-58153639 on Build GRCh38
rs868284237 Chr.20: 58153608-58153608 on Build GRCh38
rs911160415 Chr.20: 58153619-58153619 on Build GRCh38
rs772698784 Chr.20: 58153495-58153495 on Build GRCh38
rs751712742 Chr.20: 58153551-58153552 on Build GRCh38
rs756311668 Chr.20: 58153679-58153679 on Build GRCh38
rs1035202988 Chr.20: 58153510-58153510 on Build GRCh38
rs1047139164 Chr.20: 58153496-58153496 on Build GRCh38
rs111887358 Chr.20: 58153542-58153542 on Build GRCh38
rs781106052 Chr.20: 58153633-58153633 on Build GRCh38
rs923954238 Chr.20: 58153570-58153570 on Build GRCh38
rs931105104 Chr.20: 58153474-58153474 on Build GRCh38
rs373173848 Chr.20: 58153519-58153519 on Build GRCh38
rs767868075 Chr.20: 58153596-58153596 on Build GRCh38
rs747891335 Chr.20: 58153554-58153554 on Build GRCh38
rs1056191238 Chr.20: 58153666-58153666 on Build GRCh38
rs61754558 Chr.20: 58153598-58153598 on Build GRCh38
rs780117816 Chr.20: 58153545-58153545 on Build GRCh38
rs1038021451 Chr.20: 58153475-58153475 on Build GRCh38
rs371173995 Chr.20: 58153632-58153632 on Build GRCh38
rs16984945 Chr.20: 58153552-58153552 on Build GRCh38

View Full Product Details