Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 95495753-95496026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB4
Entrez Gene ID
51666
Gene Name
ankyrin repeat and SOCS box containing 4
Gene Aliases
ASB-4
UniGene
Hs.602765
Gene Chromosome Location
Chr.7: 95485439-95544738 on Build GRCh38
Transcripts
NM_016116.2 NM_145872.2 XM_017012303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6605562 c.337G>A p.V113M Chr.7: 95495907-95495907 on Build GRCh38
6297640 c.358G>T p.V120L Chr.7: 95495928-95495928 on Build GRCh38
5884609 c.233G>A p.G78D Chr.7: 95495803-95495803 on Build GRCh38
4542608 c.321G>A p.G107G Chr.7: 95495891-95495891 on Build GRCh38
1453036 c.408C>T p.Y136Y Chr.7: 95495978-95495978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758073916 Chr.7: 95495813-95495813 on Build GRCh38
rs754432763 Chr.7: 95495805-95495805 on Build GRCh38
rs777640563 Chr.7: 95495960-95495960 on Build GRCh38
rs749844932 Chr.7: 95495812-95495812 on Build GRCh38
rs547148697 Chr.7: 95495883-95495883 on Build GRCh38
rs141186023 Chr.7: 95495956-95495956 on Build GRCh38
rs114296603 Chr.7: 95495781-95495781 on Build GRCh38
rs774506865 Chr.7: 95495868-95495868 on Build GRCh38
rs539458950 Chr.7: 95495896-95495896 on Build GRCh38
rs376814974 Chr.7: 95495916-95495916 on Build GRCh38
rs762150698 Chr.7: 95495988-95495988 on Build GRCh38
rs1031541136 Chr.7: 95495799-95495799 on Build GRCh38
rs778508330 Chr.7: 95495808-95495808 on Build GRCh38
rs372932698 Chr.7: 95495961-95495961 on Build GRCh38
rs754585089 Chr.7: 95495940-95495940 on Build GRCh38
rs147091900 Chr.7: 95495981-95495981 on Build GRCh38
rs765595046 Chr.7: 95495910-95495910 on Build GRCh38
rs370663407 Chr.7: 95495917-95495917 on Build GRCh38
rs751253040 Chr.7: 95495939-95495939 on Build GRCh38
rs759469934 Chr.7: 95495873-95495873 on Build GRCh38
rs1047191434 Chr.7: 95495921-95495921 on Build GRCh38
rs765619625 Chr.7: 95495997-95495997 on Build GRCh38
rs747253499 Chr.7: 95495978-95495978 on Build GRCh38
rs761176544 Chr.7: 95495786-95495786 on Build GRCh38
rs756768517 Chr.7: 95495873-95495874 on Build GRCh38
rs762395018 Chr.7: 95495907-95495907 on Build GRCh38
rs570326706 Chr.7: 95495821-95495821 on Build GRCh38
rs779517506 Chr.7: 95495927-95495927 on Build GRCh38
rs753118507 Chr.7: 95495785-95495785 on Build GRCh38
rs769573705 Chr.7: 95495863-95495863 on Build GRCh38
rs373101368 Chr.7: 95495887-95495887 on Build GRCh38
rs746653735 Chr.7: 95495837-95495837 on Build GRCh38
rs202221772 Chr.7: 95495971-95495971 on Build GRCh38
rs757660898 Chr.7: 95495806-95495806 on Build GRCh38
rs147798502 Chr.7: 95495856-95495856 on Build GRCh38
rs759829404 Chr.7: 95495776-95495776 on Build GRCh38
rs764581157 Chr.7: 95495898-95495898 on Build GRCh38
rs757962480 Chr.7: 95495926-95495926 on Build GRCh38
rs780991883 Chr.7: 95495955-95495955 on Build GRCh38
rs748149300 Chr.7: 95495860-95495860 on Build GRCh38
rs764528090 Chr.7: 95495791-95495791 on Build GRCh38
rs768490367 Chr.7: 95495845-95495845 on Build GRCh38
rs142138079 Chr.7: 95495796-95495796 on Build GRCh38
rs777057452 Chr.7: 95495987-95495987 on Build GRCh38
rs556456967 Chr.7: 95495906-95495906 on Build GRCh38
rs1055916901 Chr.7: 95495847-95495847 on Build GRCh38

View Full Product Details