Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8228057-8228324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTC1
Entrez Gene ID
80169
Gene Name
CTS telomere maintenance complex component 1
Gene Aliases
AAF-132, AAF132, C17orf68, CRMCC, tmp494178
UniGene
Hs.156055
Gene Chromosome Location
Chr.17: 8224821-8248095 on Build GRCh38
Transcripts
NM_025099.5 NR_046431.1 XM_006721577.3 XM_006721578.3 XM_011524010.2 XM_011524011.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925921 c.3553C>T p.P1185S Chr.17: 8228281-8228281 on Build GRCh38
3712487 c.3563C>A p.T1188N Chr.17: 8228271-8228271 on Build GRCh38
986232 c.3567C>T p.H1189H Chr.17: 8228267-8228267 on Build GRCh38
319029 c.3622A>G p.S1208G Chr.17: 8228212-8228212 on Build GRCh38
390106 c.3613G>C p.E1205Q Chr.17: 8228221-8228221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938330252 Chr.17: 8228168-8228168 on Build GRCh38
rs764641685 Chr.17: 8228146-8228146 on Build GRCh38
rs751483412 Chr.17: 8228276-8228276 on Build GRCh38
rs3027245 Chr.17: 8228137-8228137 on Build GRCh38
rs1018091714 Chr.17: 8228115-8228115 on Build GRCh38
rs753366370 Chr.17: 8228131-8228131 on Build GRCh38
rs886053609 Chr.17: 8228116-8228116 on Build GRCh38
rs761178911 Chr.17: 8228234-8228234 on Build GRCh38
rs371036203 Chr.17: 8228266-8228266 on Build GRCh38
rs758988824 Chr.17: 8228106-8228106 on Build GRCh38
rs200919310 Chr.17: 8228302-8228302 on Build GRCh38
rs147714487 Chr.17: 8228230-8228230 on Build GRCh38
rs894193631 Chr.17: 8228213-8228213 on Build GRCh38
rs780090929 Chr.17: 8228158-8228158 on Build GRCh38
rs975627976 Chr.17: 8228126-8228126 on Build GRCh38
rs753200705 Chr.17: 8228223-8228224 on Build GRCh38
rs759855918 Chr.17: 8228205-8228205 on Build GRCh38
rs772166289 Chr.17: 8228233-8228233 on Build GRCh38
rs367664787 Chr.17: 8228248-8228248 on Build GRCh38
rs765992408 Chr.17: 8228274-8228274 on Build GRCh38
rs892408695 Chr.17: 8228214-8228214 on Build GRCh38
rs762508962 Chr.17: 8228271-8228271 on Build GRCh38
rs754749578 Chr.17: 8228166-8228166 on Build GRCh38
rs886053608 Chr.17: 8228115-8228115 on Build GRCh38
rs199473682 Chr.17: 8228251-8228251 on Build GRCh38
rs758386429 Chr.17: 8228153-8228153 on Build GRCh38
rs562356966 Chr.17: 8228296-8228296 on Build GRCh38
rs545385816 Chr.17: 8228108-8228108 on Build GRCh38
rs984798142 Chr.17: 8228086-8228086 on Build GRCh38
rs761493927 Chr.17: 8228177-8228177 on Build GRCh38
rs764813118 Chr.17: 8228301-8228301 on Build GRCh38
rs116523089 Chr.17: 8228109-8228109 on Build GRCh38
rs755022443 Chr.17: 8228279-8228279 on Build GRCh38
rs764093659 Chr.17: 8228275-8228275 on Build GRCh38
rs768062140 Chr.17: 8228211-8228219 on Build GRCh38
rs199473683 Chr.17: 8228228-8228248 on Build GRCh38
rs115990839 Chr.17: 8228243-8228243 on Build GRCh38
rs756625965 Chr.17: 8228267-8228267 on Build GRCh38
rs374942931 Chr.17: 8228267-8228267 on Build GRCh38
rs908765580 Chr.17: 8228090-8228090 on Build GRCh38
rs963922849 Chr.17: 8228116-8228116 on Build GRCh38
rs749988882 Chr.17: 8228149-8228149 on Build GRCh38
rs200134999 Chr.17: 8228161-8228161 on Build GRCh38
rs771253891 Chr.17: 8228222-8228222 on Build GRCh38
rs781050870 Chr.17: 8228174-8228174 on Build GRCh38
rs949148584 Chr.17: 8228129-8228129 on Build GRCh38
rs749455590 Chr.17: 8228193-8228193 on Build GRCh38
rs778830884 Chr.17: 8228229-8228229 on Build GRCh38
rs750119408 Chr.17: 8228270-8228270 on Build GRCh38
rs931194879 Chr.17: 8228148-8228148 on Build GRCh38
rs982652908 Chr.17: 8228154-8228154 on Build GRCh38

View Full Product Details