Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148827883-148828142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPB1
Entrez Gene ID
1360
Gene Name
carboxypeptidase B1
Gene Aliases
CPB, PASP, PCPB
UniGene
Hs.477891
Gene Chromosome Location
Chr.3: 148827801-148860187 on Build GRCh38
Transcripts
NM_001871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4526808 c.141G>A p.T47T Chr.3: 148828071-148828071 on Build GRCh38
6192793 c.83T>G p.F28C Chr.3: 148828013-148828013 on Build GRCh38
4525412 c.132G>A p.L44L Chr.3: 148828062-148828062 on Build GRCh38
160171 c.112A>T p.I38F Chr.3: 148828042-148828042 on Build GRCh38
1718493 c.109C>T p.H37Y Chr.3: 148828039-148828039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780393877 Chr.3: 148828102-148828103 on Build GRCh38
rs919803241 Chr.3: 148828066-148828066 on Build GRCh38
rs761076436 Chr.3: 148827953-148827953 on Build GRCh38
rs780655275 Chr.3: 148828057-148828057 on Build GRCh38
rs758577618 Chr.3: 148828056-148828056 on Build GRCh38
rs779840672 Chr.3: 148828071-148828071 on Build GRCh38
rs770895548 Chr.3: 148827907-148827907 on Build GRCh38
rs374794322 Chr.3: 148828016-148828016 on Build GRCh38
rs772648786 Chr.3: 148827936-148827936 on Build GRCh38
rs938973328 Chr.3: 148828029-148828029 on Build GRCh38
rs373628167 Chr.3: 148828015-148828015 on Build GRCh38
rs956776118 Chr.3: 148827973-148827973 on Build GRCh38
rs114058679 Chr.3: 148828054-148828054 on Build GRCh38
rs747564641 Chr.3: 148828059-148828059 on Build GRCh38
rs777675458 Chr.3: 148828024-148828024 on Build GRCh38
rs536640734 Chr.3: 148827975-148827975 on Build GRCh38
rs754000895 Chr.3: 148828025-148828025 on Build GRCh38
rs146537927 Chr.3: 148828070-148828070 on Build GRCh38
rs774986883 Chr.3: 148828094-148828094 on Build GRCh38
rs746362564 Chr.3: 148827928-148827928 on Build GRCh38
rs150860615 Chr.3: 148828003-148828003 on Build GRCh38
rs10935727 Chr.3: 148827913-148827913 on Build GRCh38
rs1021138024 Chr.3: 148827943-148827943 on Build GRCh38
rs768868275 Chr.3: 148828061-148828061 on Build GRCh38
rs759006417 Chr.3: 148828114-148828114 on Build GRCh38
rs758745005 Chr.3: 148828080-148828086 on Build GRCh38
rs193238667 Chr.3: 148828055-148828055 on Build GRCh38
rs80013498 Chr.3: 148828002-148828002 on Build GRCh38
rs558715223 Chr.3: 148827999-148827999 on Build GRCh38
rs769160991 Chr.3: 148827966-148827966 on Build GRCh38
rs199871648 Chr.3: 148828014-148828014 on Build GRCh38
rs539276743 Chr.3: 148828093-148828093 on Build GRCh38
rs757556861 Chr.3: 148828043-148828043 on Build GRCh38
rs776145474 Chr.3: 148827938-148827938 on Build GRCh38
rs773883200 Chr.3: 148828110-148828110 on Build GRCh38
rs755855538 Chr.3: 148828023-148828023 on Build GRCh38
rs772713976 Chr.3: 148827968-148827968 on Build GRCh38

View Full Product Details