Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 44977534-44977781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf10
Entrez Gene ID
11067
Gene Name
chromosome 10 open reading frame 10
Gene Aliases
DEPP, FIG, Fseg
UniGene
Hs.93675
Gene Chromosome Location
Chr.10: 44976261-44978882 on Build GRCh38
Transcripts
NM_007021.3 NM_032023.3 XM_005271833.4 XM_005271835.4 XM_006718013.3 XM_006718014.3 XM_011540255.2 XM_017016754.1 XM_017016755.1 XM_017016756.1 XM_017016757.1 XM_017016758.1

Gene Symbol
RASSF4
Entrez Gene ID
83937
Gene Name
Ras association domain family member 4
Gene Aliases
AD037
UniGene
Hs.522895
Gene Chromosome Location
Chr.10: 44959753-44994724 on Build GRCh38
Transcripts
NM_007021.3 NM_032023.3 XM_005271833.4 XM_005271835.4 XM_006718013.3 XM_006718014.3 XM_011540255.2 XM_017016754.1 XM_017016755.1 XM_017016756.1 XM_017016757.1 XM_017016758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6387183 c.375G>T p.R125S Chr.10: 44977656-44977656 on Build GRCh38
4014232 c.289C>T p.P97S Chr.10: 44977742-44977742 on Build GRCh38
1957695 c.259G>A p.A87T Chr.10: 44977772-44977772 on Build GRCh38
3415046 c.326T>G p.F109C Chr.10: 44977705-44977705 on Build GRCh38
684274 c.305T>C p.V102A Chr.10: 44977726-44977726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770431622 Chr.10: 44977558-44977558 on Build GRCh38
rs748649419 Chr.10: 44977591-44977592 on Build GRCh38
rs772159165 Chr.10: 44977619-44977623 on Build GRCh38
rs894475090 Chr.10: 44977738-44977738 on Build GRCh38
rs747217711 Chr.10: 44977627-44977628 on Build GRCh38
rs147897217 Chr.10: 44977729-44977729 on Build GRCh38
rs945476157 Chr.10: 44977669-44977669 on Build GRCh38
rs781700581 Chr.10: 44977632-44977632 on Build GRCh38
rs753621695 Chr.10: 44977592-44977592 on Build GRCh38
rs751631089 Chr.10: 44977713-44977713 on Build GRCh38
rs770794008 Chr.10: 44977611-44977611 on Build GRCh38
rs775457258 Chr.10: 44977589-44977589 on Build GRCh38
rs777781340 Chr.10: 44977625-44977626 on Build GRCh38
rs778379224 Chr.10: 44977646-44977646 on Build GRCh38
rs756486381 Chr.10: 44977741-44977741 on Build GRCh38
rs766448715 Chr.10: 44977619-44977619 on Build GRCh38
rs776762120 Chr.10: 44977594-44977594 on Build GRCh38
rs765056836 Chr.10: 44977670-44977670 on Build GRCh38
rs142587542 Chr.10: 44977595-44977595 on Build GRCh38
rs201753865 Chr.10: 44977607-44977607 on Build GRCh38
rs939888737 Chr.10: 44977682-44977682 on Build GRCh38
rs747007077 Chr.10: 44977660-44977660 on Build GRCh38
rs200209562 Chr.10: 44977726-44977727 on Build GRCh38
rs756963477 Chr.10: 44977642-44977642 on Build GRCh38
rs773914400 Chr.10: 44977566-44977566 on Build GRCh38
rs749005637 Chr.10: 44977552-44977552 on Build GRCh38
rs778171132 Chr.10: 44977743-44977743 on Build GRCh38
rs867293581 Chr.10: 44977694-44977694 on Build GRCh38
rs755524345 Chr.10: 44977626-44977626 on Build GRCh38
rs773343592 Chr.10: 44977693-44977693 on Build GRCh38
rs369762946 Chr.10: 44977623-44977623 on Build GRCh38
rs762039604 Chr.10: 44977593-44977593 on Build GRCh38
rs923106067 Chr.10: 44977661-44977661 on Build GRCh38
rs763944336 Chr.10: 44977591-44977591 on Build GRCh38
rs373182070 Chr.10: 44977637-44977637 on Build GRCh38
rs1040169078 Chr.10: 44977627-44977627 on Build GRCh38
rs758927938 Chr.10: 44977584-44977584 on Build GRCh38
rs145465141 Chr.10: 44977750-44977750 on Build GRCh38
rs776419897 Chr.10: 44977668-44977668 on Build GRCh38
rs116781265 Chr.10: 44977730-44977730 on Build GRCh38
rs774976580 Chr.10: 44977659-44977659 on Build GRCh38
rs200784305 Chr.10: 44977717-44977717 on Build GRCh38
rs771154895 Chr.10: 44977629-44977630 on Build GRCh38
rs760342416 Chr.10: 44977569-44977569 on Build GRCh38
rs763227857 Chr.10: 44977699-44977699 on Build GRCh38
rs779493781 Chr.10: 44977566-44977566 on Build GRCh38
rs766621045 Chr.10: 44977705-44977705 on Build GRCh38
rs768525833 Chr.10: 44977662-44977662 on Build GRCh38
rs986743790 Chr.10: 44977665-44977665 on Build GRCh38
rs745512890 Chr.10: 44977647-44977647 on Build GRCh38
rs377066951 Chr.10: 44977745-44977745 on Build GRCh38
rs772071380 Chr.10: 44977587-44977587 on Build GRCh38
rs565976045 Chr.10: 44977649-44977649 on Build GRCh38

View Full Product Details