Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155502941-155503202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY1
Entrez Gene ID
285888
Gene Name
canopy FGF signaling regulator 1
Gene Aliases
-
UniGene
Hs.146751
Gene Chromosome Location
Chr.7: 155501258-155533844 on Build GRCh38
Transcripts
NM_001103176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6860461 c.242-5_242-4insT p.? Chr.7: 155503109-155503110 on Build GRCh38
5145610 c.273G>A p.E91E Chr.7: 155503074-155503074 on Build GRCh38
6177006 c.278A>G p.*93W Chr.7: 155503069-155503069 on Build GRCh38
5688145 c.278A>T p.*93L Chr.7: 155503069-155503069 on Build GRCh38
745743 c.265C>A p.H89N Chr.7: 155503082-155503082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7804744 Chr.7: 155503159-155503159 on Build GRCh38
rs750200734 Chr.7: 155503110-155503111 on Build GRCh38
rs757140981 Chr.7: 155503088-155503088 on Build GRCh38
rs112187400 Chr.7: 155503084-155503084 on Build GRCh38
rs564743493 Chr.7: 155503109-155503109 on Build GRCh38
rs373207215 Chr.7: 155503061-155503061 on Build GRCh38
rs772126849 Chr.7: 155503152-155503152 on Build GRCh38
rs946983482 Chr.7: 155503127-155503127 on Build GRCh38
rs1031120950 Chr.7: 155502972-155502972 on Build GRCh38
rs774550028 Chr.7: 155503129-155503129 on Build GRCh38
rs376612451 Chr.7: 155503108-155503108 on Build GRCh38
rs780920345 Chr.7: 155503108-155503109 on Build GRCh38
rs757075378 Chr.7: 155503109-155503110 on Build GRCh38
rs201094886 Chr.7: 155503060-155503060 on Build GRCh38
rs531453308 Chr.7: 155503018-155503018 on Build GRCh38
rs748883431 Chr.7: 155503110-155503112 on Build GRCh38
rs767899263 Chr.7: 155503049-155503049 on Build GRCh38
rs779562829 Chr.7: 155503116-155503117 on Build GRCh38
rs753506856 Chr.7: 155503073-155503073 on Build GRCh38
rs768741345 Chr.7: 155503125-155503125 on Build GRCh38
rs752125701 Chr.7: 155503081-155503081 on Build GRCh38
rs78426535 Chr.7: 155503111-155503111 on Build GRCh38
rs373766528 Chr.7: 155503156-155503156 on Build GRCh38
rs377381883 Chr.7: 155503064-155503064 on Build GRCh38
rs183641482 Chr.7: 155503085-155503085 on Build GRCh38
rs376810923 Chr.7: 155503016-155503016 on Build GRCh38
rs768888943 Chr.7: 155503039-155503039 on Build GRCh38
rs774267283 Chr.7: 155503045-155503045 on Build GRCh38
rs35244674 Chr.7: 155503110-155503110 on Build GRCh38
rs926992501 Chr.7: 155503031-155503031 on Build GRCh38
rs746885961 Chr.7: 155503087-155503087 on Build GRCh38
rs1029813366 Chr.7: 155503114-155503114 on Build GRCh38
rs150737311 Chr.7: 155503166-155503166 on Build GRCh38
rs754581044 Chr.7: 155503077-155503077 on Build GRCh38
rs370495689 Chr.7: 155503146-155503146 on Build GRCh38
rs62481189 Chr.7: 155503108-155503108 on Build GRCh38
rs781096683 Chr.7: 155503101-155503101 on Build GRCh38
rs796362863 Chr.7: 155503109-155503110 on Build GRCh38
rs370137337 Chr.7: 155503135-155503135 on Build GRCh38
rs764841901 Chr.7: 155503080-155503080 on Build GRCh38
rs398006834 Chr.7: 155503123-155503123 on Build GRCh38
rs766670532 Chr.7: 155503067-155503067 on Build GRCh38
rs754709888 Chr.7: 155503048-155503048 on Build GRCh38
rs75945873 Chr.7: 155503110-155503110 on Build GRCh38

View Full Product Details