Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75105618-75105891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP13
Entrez Gene ID
51207
Gene Name
dual specificity phosphatase 13
Gene Aliases
BEDP, DUSP13A, DUSP13B, MDSP, SKRP4, TMDP
UniGene
Hs.178170
Gene Chromosome Location
Chr.10: 75094432-75109221 on Build GRCh38
Transcripts
NM_001007271.1 NM_001007272.1 NM_001007273.1 NM_001320842.1 NM_001320843.1 XM_005269883.3 XM_005269887.1 XM_005269890.1 XM_011539853.1 XM_011539854.2 XM_011539856.2 XM_017016314.1 XM_011539312.2

Gene Symbol
SAMD8
Entrez Gene ID
142891
Gene Name
sterile alpha motif domain containing 8
Gene Aliases
HEL-177, SMSr
UniGene
Hs.744986
Gene Chromosome Location
Chr.10: 75099592-75182123 on Build GRCh38
Transcripts
NM_001007271.1 NM_001007272.1 NM_001007273.1 NM_001320842.1 NM_001320843.1 XM_005269883.3 XM_005269887.1 XM_005269890.1 XM_011539853.1 XM_011539854.2 XM_011539856.2 XM_017016314.1 XM_011539312.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6401675 c.546G>A p.L182L Chr.10: 75105690-75105690 on Build GRCh38
6051985 c.548G>A p.R183Q Chr.10: 75105688-75105688 on Build GRCh38
3686867 c.555C>T p.A185A Chr.10: 75105681-75105681 on Build GRCh38
5983388 c.35C>A p.A12D Chr.10: 75105713-75105713 on Build GRCh38
3686868 c.67C>T p.R23W Chr.10: 75105681-75105681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778826754 Chr.10: 75105795-75105795 on Build GRCh38
rs918555091 Chr.10: 75105711-75105711 on Build GRCh38
rs559605382 Chr.10: 75105771-75105771 on Build GRCh38
rs867841601 Chr.10: 75105675-75105675 on Build GRCh38
rs761565456 Chr.10: 75105842-75105844 on Build GRCh38
rs373934068 Chr.10: 75105705-75105705 on Build GRCh38
rs375244324 Chr.10: 75105822-75105822 on Build GRCh38
rs927433306 Chr.10: 75105835-75105835 on Build GRCh38
rs950231175 Chr.10: 75105637-75105637 on Build GRCh38
rs1022457278 Chr.10: 75105659-75105659 on Build GRCh38
rs145460879 Chr.10: 75105823-75105823 on Build GRCh38
rs1052540973 Chr.10: 75105779-75105779 on Build GRCh38
rs370795251 Chr.10: 75105748-75105748 on Build GRCh38
rs937433669 Chr.10: 75105839-75105839 on Build GRCh38
rs901101188 Chr.10: 75105763-75105763 on Build GRCh38
rs375744952 Chr.10: 75105679-75105679 on Build GRCh38
rs201867641 Chr.10: 75105731-75105731 on Build GRCh38
rs888529301 Chr.10: 75105742-75105742 on Build GRCh38
rs529278062 Chr.10: 75105832-75105832 on Build GRCh38
rs770272452 Chr.10: 75105662-75105662 on Build GRCh38
rs151051282 Chr.10: 75105833-75105833 on Build GRCh38
rs997719084 Chr.10: 75105749-75105749 on Build GRCh38
rs773613806 Chr.10: 75105668-75105668 on Build GRCh38
rs555689681 Chr.10: 75105646-75105646 on Build GRCh38
rs879759876 Chr.10: 75105840-75105840 on Build GRCh38
rs767110628 Chr.10: 75105698-75105698 on Build GRCh38
rs989213818 Chr.10: 75105649-75105649 on Build GRCh38
rs763993036 Chr.10: 75105718-75105718 on Build GRCh38
rs766543870 Chr.10: 75105730-75105730 on Build GRCh38
rs965913587 Chr.10: 75105733-75105733 on Build GRCh38
rs374708688 Chr.10: 75105772-75105772 on Build GRCh38
rs564658792 Chr.10: 75105765-75105765 on Build GRCh38
rs541004332 Chr.10: 75105702-75105702 on Build GRCh38
rs553136425 Chr.10: 75105726-75105726 on Build GRCh38
rs201689968 Chr.10: 75105791-75105791 on Build GRCh38
rs757077281 Chr.10: 75105721-75105721 on Build GRCh38
rs942531829 Chr.10: 75105743-75105743 on Build GRCh38
rs371872854 Chr.10: 75105801-75105801 on Build GRCh38
rs141536020 Chr.10: 75105681-75105681 on Build GRCh38
rs761792119 Chr.10: 75105830-75105830 on Build GRCh38
rs774902253 Chr.10: 75105783-75105783 on Build GRCh38
rs774178209 Chr.10: 75105688-75105688 on Build GRCh38
rs374153504 Chr.10: 75105680-75105680 on Build GRCh38
rs200574063 Chr.10: 75105790-75105790 on Build GRCh38
rs753605053 Chr.10: 75105719-75105719 on Build GRCh38
rs892167321 Chr.10: 75105788-75105788 on Build GRCh38
rs767649091 Chr.10: 75105858-75105858 on Build GRCh38
rs772951962 Chr.10: 75105778-75105778 on Build GRCh38
rs368434056 Chr.10: 75105759-75105759 on Build GRCh38
rs1049170848 Chr.10: 75105854-75105854 on Build GRCh38
rs755182733 Chr.10: 75105843-75105843 on Build GRCh38
rs372813584 Chr.10: 75105761-75105761 on Build GRCh38
rs769346082 Chr.10: 75105689-75105689 on Build GRCh38
rs375361804 Chr.10: 75105757-75105757 on Build GRCh38

View Full Product Details