Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7346362-7346595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAP1
Entrez Gene ID
9744
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Gene Aliases
CENTB1
UniGene
Hs.337242
Gene Chromosome Location
Chr.17: 7336506-7354944 on Build GRCh38
Transcripts
NM_014716.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
131199 c.965A>G p.D322G Chr.17: 7346449-7346449 on Build GRCh38
3820487 c.955C>T p.L319F Chr.17: 7346439-7346439 on Build GRCh38
6148012 c.959G>T p.C320F Chr.17: 7346443-7346443 on Build GRCh38
6884978 c.933C>T p.D311D Chr.17: 7346417-7346417 on Build GRCh38
2977598 c.944G>A p.C315Y Chr.17: 7346428-7346428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368692143 Chr.17: 7346421-7346421 on Build GRCh38
rs779881575 Chr.17: 7346527-7346527 on Build GRCh38
rs778655627 Chr.17: 7346504-7346504 on Build GRCh38
rs753237133 Chr.17: 7346403-7346403 on Build GRCh38
rs79280279 Chr.17: 7346413-7346413 on Build GRCh38
rs780357304 Chr.17: 7346505-7346505 on Build GRCh38
rs956067729 Chr.17: 7346528-7346528 on Build GRCh38
rs916301882 Chr.17: 7346563-7346563 on Build GRCh38
rs758166786 Chr.17: 7346523-7346523 on Build GRCh38
rs965360589 Chr.17: 7346462-7346462 on Build GRCh38
rs997518977 Chr.17: 7346490-7346490 on Build GRCh38
rs866647736 Chr.17: 7346565-7346565 on Build GRCh38
rs914378090 Chr.17: 7346570-7346570 on Build GRCh38
rs1030376809 Chr.17: 7346508-7346508 on Build GRCh38
rs11657406 Chr.17: 7346551-7346551 on Build GRCh38
rs760010798 Chr.17: 7346395-7346395 on Build GRCh38
rs928940095 Chr.17: 7346459-7346459 on Build GRCh38
rs35144635 Chr.17: 7346401-7346401 on Build GRCh38
rs756746860 Chr.17: 7346406-7346406 on Build GRCh38
rs572149392 Chr.17: 7346423-7346423 on Build GRCh38
rs774531769 Chr.17: 7346386-7346386 on Build GRCh38
rs756808757 Chr.17: 7346461-7346461 on Build GRCh38
rs1017836179 Chr.17: 7346422-7346422 on Build GRCh38
rs542767688 Chr.17: 7346519-7346519 on Build GRCh38
rs951604108 Chr.17: 7346408-7346408 on Build GRCh38
rs745756111 Chr.17: 7346539-7346539 on Build GRCh38
rs35100940 Chr.17: 7346557-7346557 on Build GRCh38

View Full Product Details