Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26916485-26916722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP4-AS1
Entrez Gene ID
147429
Gene Name
AQP4 antisense RNA 1
Gene Aliases
C18orf16, CHST9-AS1
UniGene
Hs.559280
Gene Chromosome Location
Chr.18: 26865308-26935946 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_005258362.3 XM_017026033.1 XM_017026034.1

Gene Symbol
CHST9
Entrez Gene ID
83539
Gene Name
carbohydrate sulfotransferase 9
Gene Aliases
GALNAC4ST-2, GalNAc4ST2
UniGene
Hs.44584
Gene Chromosome Location
Chr.18: 26908754-27185364 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_005258362.3 XM_017026033.1 XM_017026034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5717960 c.940C>T p.R314* Chr.18: 26916651-26916651 on Build GRCh38
708219 c.681A>T p.K227N Chr.18: 26916655-26916655 on Build GRCh38
5565426 c.618G>T p.R206S Chr.18: 26916718-26916718 on Build GRCh38
6990506 c.828G>C p.L276F Chr.18: 26916508-26916508 on Build GRCh38
2808265 c.915C>T p.F305F Chr.18: 26916676-26916676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774642167 Chr.18: 26916653-26916653 on Build GRCh38
rs767048741 Chr.18: 26916669-26916669 on Build GRCh38
rs762059727 Chr.18: 26916527-26916527 on Build GRCh38
rs780558150 Chr.18: 26916641-26916641 on Build GRCh38
rs968829896 Chr.18: 26916675-26916675 on Build GRCh38
rs200393307 Chr.18: 26916531-26916531 on Build GRCh38
rs777470513 Chr.18: 26916567-26916567 on Build GRCh38
rs773425949 Chr.18: 26916673-26916673 on Build GRCh38
rs760755840 Chr.18: 26916541-26916541 on Build GRCh38
rs201523792 Chr.18: 26916650-26916650 on Build GRCh38
rs781009083 Chr.18: 26916604-26916604 on Build GRCh38
rs769029082 Chr.18: 26916651-26916651 on Build GRCh38
rs544368300 Chr.18: 26916693-26916693 on Build GRCh38
rs1003144810 Chr.18: 26916595-26916595 on Build GRCh38
rs376394477 Chr.18: 26916545-26916545 on Build GRCh38
rs369998401 Chr.18: 26916566-26916566 on Build GRCh38
rs867199551 Chr.18: 26916553-26916553 on Build GRCh38
rs757033006 Chr.18: 26916586-26916586 on Build GRCh38
rs1054450244 Chr.18: 26916581-26916581 on Build GRCh38
rs970673625 Chr.18: 26916695-26916695 on Build GRCh38
rs377190620 Chr.18: 26916676-26916676 on Build GRCh38
rs752600019 Chr.18: 26916557-26916557 on Build GRCh38
rs750559823 Chr.18: 26916535-26916535 on Build GRCh38
rs762008758 Chr.18: 26916655-26916655 on Build GRCh38
rs533390598 Chr.18: 26916514-26916514 on Build GRCh38
rs1010251770 Chr.18: 26916668-26916668 on Build GRCh38
rs751254187 Chr.18: 26916577-26916577 on Build GRCh38
rs765154888 Chr.18: 26916543-26916543 on Build GRCh38
rs576654934 Chr.18: 26916615-26916615 on Build GRCh38
rs755666392 Chr.18: 26916598-26916598 on Build GRCh38
rs1047731634 Chr.18: 26916515-26916515 on Build GRCh38
rs745315555 Chr.18: 26916605-26916605 on Build GRCh38

View Full Product Details