Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122206764-122207033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT4
Entrez Gene ID
79369
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Gene Aliases
B3GN-T4, beta3Gn-T4
UniGene
Hs.363315
Gene Chromosome Location
Chr.12: 122203681-122208135 on Build GRCh38
Transcripts
NM_030765.2 XM_011538732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7314657 c.682G>T p.V228L Chr.12: 122206933-122206933 on Build GRCh38
1184318 c.725T>C p.L242P Chr.12: 122206976-122206976 on Build GRCh38
3968043 c.562G>A p.E188K Chr.12: 122206813-122206813 on Build GRCh38
1184319 c.679A>G p.N227D Chr.12: 122206930-122206930 on Build GRCh38
116246 c.538G>A p.D180N Chr.12: 122206789-122206789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752837963 Chr.12: 122206946-122206946 on Build GRCh38
rs768800797 Chr.12: 122206791-122206791 on Build GRCh38
rs553617934 Chr.12: 122206906-122206906 on Build GRCh38
rs187124215 Chr.12: 122206818-122206818 on Build GRCh38
rs774168455 Chr.12: 122206824-122206824 on Build GRCh38
rs966234662 Chr.12: 122206974-122206974 on Build GRCh38
rs748471148 Chr.12: 122206854-122206854 on Build GRCh38
rs897573945 Chr.12: 122206873-122206873 on Build GRCh38
rs779055570 Chr.12: 122206901-122206901 on Build GRCh38
rs956743677 Chr.12: 122207003-122207003 on Build GRCh38
rs753888870 Chr.12: 122206891-122206891 on Build GRCh38
rs199538224 Chr.12: 122206935-122206935 on Build GRCh38
rs145634810 Chr.12: 122206933-122206933 on Build GRCh38
rs369076213 Chr.12: 122206832-122206832 on Build GRCh38
rs756056900 Chr.12: 122206836-122206836 on Build GRCh38
rs765268501 Chr.12: 122206940-122206940 on Build GRCh38
rs757917533 Chr.12: 122206952-122206962 on Build GRCh38
rs760658090 Chr.12: 122206886-122206886 on Build GRCh38
rs777981592 Chr.12: 122206959-122206959 on Build GRCh38
rs965409107 Chr.12: 122206937-122206937 on Build GRCh38
rs761654592 Chr.12: 122206875-122206875 on Build GRCh38
rs762387783 Chr.12: 122206870-122206870 on Build GRCh38
rs776974249 Chr.12: 122206886-122206886 on Build GRCh38
rs767399145 Chr.12: 122206880-122206880 on Build GRCh38
rs775048350 Chr.12: 122206861-122206861 on Build GRCh38
rs777417308 Chr.12: 122206859-122206859 on Build GRCh38
rs773127810 Chr.12: 122206932-122206932 on Build GRCh38
rs375787732 Chr.12: 122206848-122206848 on Build GRCh38
rs576192434 Chr.12: 122206969-122206969 on Build GRCh38
rs138745438 Chr.12: 122206896-122206896 on Build GRCh38
rs748525931 Chr.12: 122206805-122206805 on Build GRCh38
rs766286373 Chr.12: 122206889-122206889 on Build GRCh38
rs771404601 Chr.12: 122206858-122206858 on Build GRCh38
rs762658951 Chr.12: 122206820-122206820 on Build GRCh38
rs370749355 Chr.12: 122206985-122206985 on Build GRCh38
rs201025149 Chr.12: 122206834-122206834 on Build GRCh38
rs774899917 Chr.12: 122206819-122206819 on Build GRCh38
rs373879514 Chr.12: 122206908-122206908 on Build GRCh38
rs944132769 Chr.12: 122206806-122206806 on Build GRCh38
rs767112200 Chr.12: 122206955-122206955 on Build GRCh38
rs748550610 Chr.12: 122206908-122206909 on Build GRCh38
rs745507521 Chr.12: 122206937-122206938 on Build GRCh38
rs377393964 Chr.12: 122206899-122206899 on Build GRCh38
rs369992469 Chr.12: 122206884-122206884 on Build GRCh38
rs1056583190 Chr.12: 122206929-122206929 on Build GRCh38
rs543502597 Chr.12: 122206980-122206980 on Build GRCh38
rs140444368 Chr.12: 122206924-122206924 on Build GRCh38
rs779112409 Chr.12: 122206853-122206853 on Build GRCh38
rs758995947 Chr.12: 122206815-122206815 on Build GRCh38
rs59453864 Chr.12: 122206994-122206994 on Build GRCh38
rs888718833 Chr.12: 122206837-122206837 on Build GRCh38
rs757597567 Chr.12: 122206967-122206967 on Build GRCh38
rs746769802 Chr.12: 122206993-122206993 on Build GRCh38
rs866253298 Chr.12: 122206936-122206937 on Build GRCh38
rs770153583 Chr.12: 122206911-122206928 on Build GRCh38
rs147657286 Chr.12: 122206881-122206881 on Build GRCh38
rs773738674 Chr.12: 122206811-122206811 on Build GRCh38
rs772500588 Chr.12: 122206808-122206808 on Build GRCh38
rs771793938 Chr.12: 122206992-122206992 on Build GRCh38
rs751346265 Chr.12: 122206829-122206829 on Build GRCh38
rs199695726 Chr.12: 122206857-122206857 on Build GRCh38
rs749396322 Chr.12: 122206840-122206840 on Build GRCh38
rs781558278 Chr.12: 122206923-122206923 on Build GRCh38
rs747386259 Chr.12: 122206918-122206918 on Build GRCh38
rs769430362 Chr.12: 122206818-122206820 on Build GRCh38
rs775335390 Chr.12: 122206866-122206866 on Build GRCh38
rs759565227 Chr.12: 122206939-122206939 on Build GRCh38
rs766494063 Chr.12: 122206955-122206983 on Build GRCh38
rs758601645 Chr.12: 122206950-122206950 on Build GRCh38
rs138266947 Chr.12: 122206986-122206986 on Build GRCh38
rs200448613 Chr.12: 122206921-122206921 on Build GRCh38
rs751887736 Chr.12: 122206963-122206963 on Build GRCh38
rs1002552024 Chr.12: 122207000-122207000 on Build GRCh38
rs778158051 Chr.12: 122206856-122206856 on Build GRCh38
rs760059825 Chr.12: 122206813-122206815 on Build GRCh38
rs774774427 Chr.12: 122206792-122206792 on Build GRCh38
rs768355359 Chr.12: 122206874-122206875 on Build GRCh38
rs780153302 Chr.12: 122206839-122206839 on Build GRCh38
rs951972920 Chr.12: 122206957-122206957 on Build GRCh38
rs376839421 Chr.12: 122206968-122206968 on Build GRCh38
rs753741736 Chr.12: 122206938-122206938 on Build GRCh38
rs1057256645 Chr.12: 122207005-122207005 on Build GRCh38
rs772447869 Chr.12: 122206855-122206855 on Build GRCh38
rs774180580 Chr.12: 122206874-122206874 on Build GRCh38
rs746674091 Chr.12: 122206855-122206856 on Build GRCh38
rs761527942 Chr.12: 122206927-122206927 on Build GRCh38
rs778104699 Chr.12: 122206909-122206909 on Build GRCh38

View Full Product Details