Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137049725-137049960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884817 c.1105C>A p.L369M Chr.9: 137049914-137049914 on Build GRCh38
6259272 c.1095C>A p.T365T Chr.9: 137049924-137049924 on Build GRCh38
1461305 c.1100A>G p.Q367R Chr.9: 137049919-137049919 on Build GRCh38
6550620 c.1110G>A p.E370E Chr.9: 137049909-137049909 on Build GRCh38
4681047 c.1064delT p.L355fs*20 Chr.9: 137049955-137049955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773432838 Chr.9: 137049926-137049926 on Build GRCh38
rs369266590 Chr.9: 137049940-137049940 on Build GRCh38
rs756968721 Chr.9: 137049898-137049898 on Build GRCh38
rs980923213 Chr.9: 137049753-137049753 on Build GRCh38
rs753234356 Chr.9: 137049867-137049867 on Build GRCh38
rs777276731 Chr.9: 137049885-137049885 on Build GRCh38
rs780851799 Chr.9: 137049821-137049821 on Build GRCh38
rs749270902 Chr.9: 137049939-137049939 on Build GRCh38
rs905570321 Chr.9: 137049814-137049814 on Build GRCh38
rs749263674 Chr.9: 137049916-137049916 on Build GRCh38
rs146220439 Chr.9: 137049803-137049803 on Build GRCh38
rs895737169 Chr.9: 137049820-137049820 on Build GRCh38
rs751002470 Chr.9: 137049892-137049892 on Build GRCh38
rs1043646778 Chr.9: 137049786-137049786 on Build GRCh38
rs139163179 Chr.9: 137049829-137049829 on Build GRCh38
rs988446891 Chr.9: 137049900-137049900 on Build GRCh38
rs17853461 Chr.9: 137049909-137049909 on Build GRCh38
rs947035894 Chr.9: 137049799-137049799 on Build GRCh38
rs532341072 Chr.9: 137049813-137049813 on Build GRCh38
rs756253482 Chr.9: 137049889-137049892 on Build GRCh38
rs973487149 Chr.9: 137049756-137049756 on Build GRCh38
rs553876051 Chr.9: 137049793-137049793 on Build GRCh38
rs369943432 Chr.9: 137049866-137049866 on Build GRCh38
rs374344954 Chr.9: 137049849-137049849 on Build GRCh38
rs1001375521 Chr.9: 137049817-137049817 on Build GRCh38
rs554522647 Chr.9: 137049758-137049758 on Build GRCh38
rs45526831 Chr.9: 137049860-137049860 on Build GRCh38
rs758852994 Chr.9: 137049880-137049880 on Build GRCh38
rs560275916 Chr.9: 137049779-137049779 on Build GRCh38
rs754223882 Chr.9: 137049861-137049861 on Build GRCh38
rs538152951 Chr.9: 137049757-137049757 on Build GRCh38
rs759932657 Chr.9: 137049863-137049863 on Build GRCh38
rs935574355 Chr.9: 137049778-137049778 on Build GRCh38
rs768660044 Chr.9: 137049925-137049925 on Build GRCh38
rs760994334 Chr.9: 137049854-137049854 on Build GRCh38
rs200426486 Chr.9: 137049889-137049889 on Build GRCh38
rs932155291 Chr.9: 137049760-137049760 on Build GRCh38
rs912087571 Chr.9: 137049877-137049877 on Build GRCh38
rs111660431 Chr.9: 137049769-137049769 on Build GRCh38
rs377411787 Chr.9: 137049933-137049933 on Build GRCh38
rs545856950 Chr.9: 137049811-137049811 on Build GRCh38
rs371960899 Chr.9: 137049837-137049837 on Build GRCh38
rs374124039 Chr.9: 137049826-137049826 on Build GRCh38
rs375472167 Chr.9: 137049865-137049865 on Build GRCh38
rs779678618 Chr.9: 137049914-137049914 on Build GRCh38
rs780206628 Chr.9: 137049914-137049914 on Build GRCh38
rs775472027 Chr.9: 137049836-137049836 on Build GRCh38
rs540213991 Chr.9: 137049932-137049932 on Build GRCh38
rs1056883865 Chr.9: 137049938-137049938 on Build GRCh38
rs746059138 Chr.9: 137049827-137049839 on Build GRCh38
rs376669519 Chr.9: 137049902-137049902 on Build GRCh38
rs771118429 Chr.9: 137049931-137049931 on Build GRCh38
rs769673923 Chr.9: 137049831-137049831 on Build GRCh38
rs1015242298 Chr.9: 137049832-137049832 on Build GRCh38
rs148098204 Chr.9: 137049911-137049911 on Build GRCh38
rs1015592849 Chr.9: 137049843-137049843 on Build GRCh38
rs773379719 Chr.9: 137049852-137049852 on Build GRCh38
rs770510962 Chr.9: 137049819-137049819 on Build GRCh38
rs963690275 Chr.9: 137049750-137049750 on Build GRCh38
rs771892797 Chr.9: 137049927-137049927 on Build GRCh38
rs533923881 Chr.9: 137049770-137049770 on Build GRCh38
rs913549746 Chr.9: 137049934-137049934 on Build GRCh38
rs373079181 Chr.9: 137049921-137049921 on Build GRCh38
rs1051578098 Chr.9: 137049807-137049807 on Build GRCh38

View Full Product Details