Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 108214402-108214671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3E
Entrez Gene ID
3646
Gene Name
eukaryotic translation initiation factor 3 subunit E
Gene Aliases
EIF3-P48, EIF3S6, INT6, eIF3-p46
UniGene
Hs.405590
Gene Chromosome Location
Chr.8: 108201743-108248802 on Build GRCh38
Transcripts
NM_001568.2 XM_017013398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3271542 c.1032C>G p.F344L Chr.8: 108214636-108214636 on Build GRCh38
1187602 c.1026G>C p.E342D Chr.8: 108214642-108214642 on Build GRCh38
377310 c.1032C>T p.F344F Chr.8: 108214636-108214636 on Build GRCh38
6027563 c.1007C>T p.A336V Chr.8: 108214661-108214661 on Build GRCh38
3271544 c.1010G>A p.R337H Chr.8: 108214658-108214658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773346324 Chr.8: 108214631-108214631 on Build GRCh38
rs745513411 Chr.8: 108214582-108214582 on Build GRCh38
rs769629413 Chr.8: 108214594-108214594 on Build GRCh38
rs756704478 Chr.8: 108214574-108214574 on Build GRCh38
rs376537156 Chr.8: 108214595-108214595 on Build GRCh38
rs761858354 Chr.8: 108214625-108214625 on Build GRCh38
rs531254487 Chr.8: 108214430-108214430 on Build GRCh38
rs901136062 Chr.8: 108214535-108214535 on Build GRCh38
rs989163197 Chr.8: 108214604-108214604 on Build GRCh38
rs151050498 Chr.8: 108214477-108214477 on Build GRCh38
rs749590611 Chr.8: 108214569-108214570 on Build GRCh38
rs368520339 Chr.8: 108214596-108214596 on Build GRCh38
rs536892095 Chr.8: 108214554-108214554 on Build GRCh38
rs768599555 Chr.8: 108214600-108214600 on Build GRCh38
rs993983672 Chr.8: 108214434-108214434 on Build GRCh38
rs1039824328 Chr.8: 108214482-108214482 on Build GRCh38
rs954507355 Chr.8: 108214470-108214470 on Build GRCh38
rs567835555 Chr.8: 108214537-108214537 on Build GRCh38
rs760870444 Chr.8: 108214632-108214632 on Build GRCh38
rs1026188923 Chr.8: 108214436-108214436 on Build GRCh38
rs780892229 Chr.8: 108214581-108214581 on Build GRCh38
rs769102507 Chr.8: 108214587-108214597 on Build GRCh38
rs199858643 Chr.8: 108214612-108214612 on Build GRCh38
rs772011864 Chr.8: 108214627-108214627 on Build GRCh38
rs1022067524 Chr.8: 108214641-108214641 on Build GRCh38

View Full Product Details