Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1785841-1786114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOR2
Entrez Gene ID
79602
Gene Name
adiponectin receptor 2
Gene Aliases
ACDCR2, PAQR2
UniGene
Hs.371642
Gene Chromosome Location
Chr.12: 1690981-1788679 on Build GRCh38
Transcripts
NM_024551.2 XM_005253789.1 XM_006719018.2 XM_011521024.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3739541 c.1126A>T p.I376F Chr.12: 1786037-1786037 on Build GRCh38
5027515 c.1036C>T p.H346Y Chr.12: 1785947-1785947 on Build GRCh38
6267062 c.1063G>T p.V355L Chr.12: 1785974-1785974 on Build GRCh38
937747 c.1118G>A p.R373H Chr.12: 1786029-1786029 on Build GRCh38
6462467 c.1131C>G p.G377G Chr.12: 1786042-1786042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777002870 Chr.12: 1785928-1785929 on Build GRCh38
rs779587593 Chr.12: 1785928-1785928 on Build GRCh38
rs748514165 Chr.12: 1785934-1785935 on Build GRCh38
rs768303586 Chr.12: 1785938-1785938 on Build GRCh38
rs550948279 Chr.12: 1785887-1785887 on Build GRCh38
rs770166457 Chr.12: 1786078-1786081 on Build GRCh38
rs573669927 Chr.12: 1786080-1786080 on Build GRCh38
rs896260025 Chr.12: 1785964-1785964 on Build GRCh38
rs760687680 Chr.12: 1786000-1786000 on Build GRCh38
rs768828151 Chr.12: 1785924-1785926 on Build GRCh38
rs377162169 Chr.12: 1785925-1785925 on Build GRCh38
rs768176210 Chr.12: 1785986-1785986 on Build GRCh38
rs747236043 Chr.12: 1785916-1785916 on Build GRCh38
rs141185371 Chr.12: 1786009-1786009 on Build GRCh38
rs760094751 Chr.12: 1785967-1785967 on Build GRCh38
rs149429128 Chr.12: 1785903-1785903 on Build GRCh38
rs747784888 Chr.12: 1786065-1786065 on Build GRCh38
rs767689143 Chr.12: 1785893-1785893 on Build GRCh38
rs778658717 Chr.12: 1786033-1786033 on Build GRCh38
rs374984352 Chr.12: 1786039-1786039 on Build GRCh38
rs376869220 Chr.12: 1786079-1786079 on Build GRCh38
rs752776070 Chr.12: 1785997-1785997 on Build GRCh38
rs138917773 Chr.12: 1785869-1785869 on Build GRCh38
rs774822754 Chr.12: 1786069-1786069 on Build GRCh38
rs777935318 Chr.12: 1785966-1785966 on Build GRCh38
rs768056305 Chr.12: 1786062-1786062 on Build GRCh38
rs773664032 Chr.12: 1786081-1786082 on Build GRCh38
rs760004494 Chr.12: 1786074-1786074 on Build GRCh38
rs529232610 Chr.12: 1785878-1785878 on Build GRCh38
rs774910578 Chr.12: 1785958-1785958 on Build GRCh38
rs730031 Chr.12: 1786042-1786042 on Build GRCh38
rs954028523 Chr.12: 1786082-1786082 on Build GRCh38
rs201876183 Chr.12: 1785907-1785907 on Build GRCh38
rs779876961 Chr.12: 1786048-1786048 on Build GRCh38
rs753615186 Chr.12: 1785919-1785919 on Build GRCh38
rs767407914 Chr.12: 1786055-1786055 on Build GRCh38
rs757534843 Chr.12: 1786032-1786032 on Build GRCh38
rs763310434 Chr.12: 1785946-1785946 on Build GRCh38
rs746469299 Chr.12: 1786061-1786061 on Build GRCh38
rs761314663 Chr.12: 1786081-1786081 on Build GRCh38
rs777883690 Chr.12: 1785913-1785913 on Build GRCh38
rs1000788514 Chr.12: 1786077-1786077 on Build GRCh38
rs201089342 Chr.12: 1785931-1785931 on Build GRCh38
rs905386371 Chr.12: 1785886-1785886 on Build GRCh38
rs139431821 Chr.12: 1786064-1786064 on Build GRCh38
rs757053727 Chr.12: 1785920-1785920 on Build GRCh38
rs746529758 Chr.12: 1785935-1785935 on Build GRCh38
rs745791984 Chr.12: 1785926-1785926 on Build GRCh38
rs769616920 Chr.12: 1786066-1786066 on Build GRCh38
rs754002919 Chr.12: 1786029-1786029 on Build GRCh38
rs372797102 Chr.12: 1785927-1785927 on Build GRCh38
rs776161287 Chr.12: 1785941-1785941 on Build GRCh38

View Full Product Details