Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31189240-31189513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN14
Entrez Gene ID
440854
Gene Name
calpain 14
Gene Aliases
-
UniGene
Hs.468059
Gene Chromosome Location
Chr.2: 31173056-31233957 on Build GRCh38
Transcripts
NM_001145122.1 NM_001321270.1 XM_011532863.2 XM_011532864.2 XM_011532865.1 XM_011532866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4619532 c.1382G>T p.S461I Chr.2: 31189384-31189384 on Build GRCh38
6434759 c.1367A>G p.K456R Chr.2: 31189399-31189399 on Build GRCh38
6432160 c.1402C>A p.P468T Chr.2: 31189364-31189364 on Build GRCh38
5599237 c.1475C>T p.S492F Chr.2: 31189291-31189291 on Build GRCh38
5396901 c.1369G>A p.E457K Chr.2: 31189397-31189397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759569414 Chr.2: 31189272-31189272 on Build GRCh38
rs1014056479 Chr.2: 31189381-31189381 on Build GRCh38
rs750442499 Chr.2: 31189451-31189451 on Build GRCh38
rs750697717 Chr.2: 31189452-31189452 on Build GRCh38
rs370449152 Chr.2: 31189325-31189325 on Build GRCh38
rs745984537 Chr.2: 31189493-31189493 on Build GRCh38
rs145387315 Chr.2: 31189392-31189392 on Build GRCh38
rs775493753 Chr.2: 31189332-31189332 on Build GRCh38
rs758388235 Chr.2: 31189455-31189455 on Build GRCh38
rs192034616 Chr.2: 31189401-31189401 on Build GRCh38
rs780665948 Chr.2: 31189282-31189282 on Build GRCh38
rs774839785 Chr.2: 31189319-31189319 on Build GRCh38
rs753959703 Chr.2: 31189357-31189357 on Build GRCh38
rs934342908 Chr.2: 31189361-31189361 on Build GRCh38
rs377236419 Chr.2: 31189344-31189344 on Build GRCh38
rs985596200 Chr.2: 31189302-31189302 on Build GRCh38
rs764875851 Chr.2: 31189426-31189426 on Build GRCh38
rs1017016137 Chr.2: 31189478-31189478 on Build GRCh38
rs1022303100 Chr.2: 31189338-31189338 on Build GRCh38
rs769608402 Chr.2: 31189307-31189307 on Build GRCh38
rs113806756 Chr.2: 31189308-31189308 on Build GRCh38
rs752357052 Chr.2: 31189335-31189335 on Build GRCh38
rs999285869 Chr.2: 31189356-31189356 on Build GRCh38
rs895148222 Chr.2: 31189313-31189313 on Build GRCh38
rs201587590 Chr.2: 31189346-31189346 on Build GRCh38
rs370557764 Chr.2: 31189371-31189371 on Build GRCh38
rs1053448992 Chr.2: 31189363-31189363 on Build GRCh38
rs763923309 Chr.2: 31189418-31189418 on Build GRCh38
rs78099670 Chr.2: 31189481-31189481 on Build GRCh38
rs905690667 Chr.2: 31189450-31189450 on Build GRCh38
rs199725219 Chr.2: 31189343-31189343 on Build GRCh38
rs551754163 Chr.2: 31189432-31189432 on Build GRCh38
rs368597382 Chr.2: 31189347-31189347 on Build GRCh38

View Full Product Details