Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 46574010-46574237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST7
Entrez Gene ID
56548
Gene Name
carbohydrate sulfotransferase 7
Gene Aliases
C6ST-2, GST-5
UniGene
Hs.129955
Gene Chromosome Location
Chr.X: 46573687-46598496 on Build GRCh38
Transcripts
NM_019886.3

SNP Detail

SNP ID Chromosome Location
rs781382088 Chr.X: 46574210-46574210 on Build GRCh38
rs777555910 Chr.X: 46574161-46574161 on Build GRCh38
rs1057072866 Chr.X: 46574121-46574121 on Build GRCh38
rs773167607 Chr.X: 46574057-46574057 on Build GRCh38
rs770884974 Chr.X: 46574217-46574217 on Build GRCh38
rs1006895013 Chr.X: 46574179-46574179 on Build GRCh38
rs759196964 Chr.X: 46574117-46574117 on Build GRCh38
rs773884876 Chr.X: 46574091-46574091 on Build GRCh38
rs767273988 Chr.X: 46574120-46574120 on Build GRCh38
rs771837285 Chr.X: 46574042-46574042 on Build GRCh38
rs765705791 Chr.X: 46574150-46574150 on Build GRCh38
rs369680575 Chr.X: 46574144-46574144 on Build GRCh38
rs751788961 Chr.X: 46574188-46574188 on Build GRCh38
rs769680098 Chr.X: 46574055-46574055 on Build GRCh38
rs762398181 Chr.X: 46574058-46574058 on Build GRCh38
rs754310332 Chr.X: 46574134-46574134 on Build GRCh38
rs747935476 Chr.X: 46574035-46574035 on Build GRCh38
rs200180964 Chr.X: 46574201-46574201 on Build GRCh38
rs1023191728 Chr.X: 46574211-46574211 on Build GRCh38
rs755895426 Chr.X: 46574213-46574213 on Build GRCh38
rs770422083 Chr.X: 46574068-46574068 on Build GRCh38
rs895785535 Chr.X: 46574140-46574140 on Build GRCh38
rs781021631 Chr.X: 46574109-46574128 on Build GRCh38
rs766424462 Chr.X: 46574182-46574182 on Build GRCh38
rs750903362 Chr.X: 46574160-46574160 on Build GRCh38
rs904627226 Chr.X: 46574181-46574181 on Build GRCh38
rs780902051 Chr.X: 46574032-46574032 on Build GRCh38
rs1031871173 Chr.X: 46574207-46574207 on Build GRCh38

View Full Product Details