Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 54816714-54816953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML6
Entrez Gene ID
400954
Gene Name
echinoderm microtubule associated protein like 6
Gene Aliases
-
UniGene
Hs.656692
Gene Chromosome Location
Chr.2: 54723467-54972908 on Build GRCh38
Transcripts
NM_001039753.2 XM_017004098.1 XM_017004099.1 XM_017004100.1 XM_017004102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7280299 c.375G>A p.G125G Chr.2: 54816804-54816804 on Build GRCh38
1205278 c.407A>T p.D136V Chr.2: 54816836-54816836 on Build GRCh38
6226580 c.432G>A p.A144A Chr.2: 54816861-54816861 on Build GRCh38
1408584 c.413G>T p.R138M Chr.2: 54816842-54816842 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373616236 Chr.2: 54816893-54816893 on Build GRCh38
rs775581352 Chr.2: 54816882-54816882 on Build GRCh38
rs769549311 Chr.2: 54816876-54816876 on Build GRCh38
rs965422561 Chr.2: 54816779-54816779 on Build GRCh38
rs1028212277 Chr.2: 54816788-54816788 on Build GRCh38
rs755360982 Chr.2: 54816821-54816821 on Build GRCh38
rs765100524 Chr.2: 54816921-54816921 on Build GRCh38
rs993803235 Chr.2: 54816754-54816767 on Build GRCh38
rs970823693 Chr.2: 54816798-54816799 on Build GRCh38
rs942902529 Chr.2: 54816875-54816875 on Build GRCh38
rs1046660412 Chr.2: 54816869-54816869 on Build GRCh38
rs953989776 Chr.2: 54816804-54816804 on Build GRCh38
rs186281872 Chr.2: 54816870-54816870 on Build GRCh38
rs1032183231 Chr.2: 54816746-54816746 on Build GRCh38
rs754147048 Chr.2: 54816784-54816784 on Build GRCh38
rs766916949 Chr.2: 54816776-54816776 on Build GRCh38
rs761940801 Chr.2: 54816751-54816751 on Build GRCh38
rs952966555 Chr.2: 54816842-54816842 on Build GRCh38
rs749042204 Chr.2: 54816902-54816902 on Build GRCh38
rs541784728 Chr.2: 54816871-54816871 on Build GRCh38
rs767530733 Chr.2: 54816756-54816756 on Build GRCh38
rs900810780 Chr.2: 54816925-54816925 on Build GRCh38
rs763718947 Chr.2: 54816846-54816846 on Build GRCh38
rs370983931 Chr.2: 54816914-54816914 on Build GRCh38
rs756771011 Chr.2: 54816847-54816847 on Build GRCh38
rs750917481 Chr.2: 54816760-54816760 on Build GRCh38
rs919231664 Chr.2: 54816856-54816856 on Build GRCh38
rs377645256 Chr.2: 54816837-54816837 on Build GRCh38
rs369858624 Chr.2: 54816822-54816822 on Build GRCh38
rs370463321 Chr.2: 54816873-54816873 on Build GRCh38
rs374066595 Chr.2: 54816834-54816834 on Build GRCh38
rs545074589 Chr.2: 54816767-54816767 on Build GRCh38
rs911470163 Chr.2: 54816866-54816866 on Build GRCh38
rs768602897 Chr.2: 54816909-54816909 on Build GRCh38
rs949427800 Chr.2: 54816858-54816858 on Build GRCh38
rs780413903 Chr.2: 54816893-54816894 on Build GRCh38

View Full Product Details