Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3191085-3191348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDRGK1
Entrez Gene ID
65992
Gene Name
DDRGK domain containing 1
Gene Aliases
C20orf116, UFBP1, dJ1187M17.3
UniGene
Hs.471975
Gene Chromosome Location
Chr.20: 3190350-3204697 on Build GRCh38
Transcripts
NM_023935.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025877 c.750G>T p.Q250H Chr.20: 3191218-3191218 on Build GRCh38
4801056 c.736A>G p.I246V Chr.20: 3191232-3191232 on Build GRCh38
3545397 c.754C>T p.L252L Chr.20: 3191214-3191214 on Build GRCh38
4918575 c.741T>A p.N247K Chr.20: 3191227-3191227 on Build GRCh38
7363945 c.777A>T p.T259T Chr.20: 3191191-3191191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751631925 Chr.20: 3191172-3191172 on Build GRCh38
rs770146758 Chr.20: 3191225-3191225 on Build GRCh38
rs748073184 Chr.20: 3191269-3191269 on Build GRCh38
rs746048840 Chr.20: 3191208-3191208 on Build GRCh38
rs761289201 Chr.20: 3191248-3191248 on Build GRCh38
rs368305863 Chr.20: 3191171-3191171 on Build GRCh38
rs959761663 Chr.20: 3191276-3191276 on Build GRCh38
rs769200812 Chr.20: 3191245-3191245 on Build GRCh38
rs943882277 Chr.20: 3191133-3191134 on Build GRCh38
rs370418437 Chr.20: 3191254-3191254 on Build GRCh38
rs934585432 Chr.20: 3191241-3191241 on Build GRCh38
rs1043710536 Chr.20: 3191209-3191209 on Build GRCh38
rs555131014 Chr.20: 3191141-3191141 on Build GRCh38
rs757463636 Chr.20: 3191179-3191179 on Build GRCh38
rs778980641 Chr.20: 3191119-3191119 on Build GRCh38
rs371714511 Chr.20: 3191309-3191309 on Build GRCh38
rs772787330 Chr.20: 3191199-3191200 on Build GRCh38
rs536785558 Chr.20: 3191139-3191139 on Build GRCh38
rs760251165 Chr.20: 3191278-3191278 on Build GRCh38
rs749674219 Chr.20: 3191232-3191232 on Build GRCh38
rs151280429 Chr.20: 3191226-3191226 on Build GRCh38
rs1048756217 Chr.20: 3191213-3191213 on Build GRCh38
rs781446482 Chr.20: 3191196-3191196 on Build GRCh38
rs11906805 Chr.20: 3191285-3191285 on Build GRCh38
rs761085623 Chr.20: 3191243-3191245 on Build GRCh38
rs979711113 Chr.20: 3191305-3191305 on Build GRCh38
rs1039540678 Chr.20: 3191142-3191142 on Build GRCh38
rs774760760 Chr.20: 3191246-3191246 on Build GRCh38

View Full Product Details