Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203219619-203219890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIT1
Entrez Gene ID
1118
Gene Name
chitinase 1
Gene Aliases
CHI3, CHIT, CHITD
UniGene
Hs.201688
Gene Chromosome Location
Chr.1: 203216079-203229732 on Build GRCh38
Transcripts
NM_001256125.1 NM_001270509.1 NM_003465.2 NR_045784.1 NR_045785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1733798 c.730-2A>G p.? Chr.1: 203219851-203219851 on Build GRCh38
5914585 c.884C>T p.T295I Chr.1: 203219695-203219695 on Build GRCh38
6368211 c.755T>C p.I252T Chr.1: 203219797-203219797 on Build GRCh38
463778 c.833A>T p.D278V Chr.1: 203219746-203219746 on Build GRCh38
5901014 c.839G>A p.R280K Chr.1: 203219740-203219740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774345722 Chr.1: 203219825-203219825 on Build GRCh38
rs759317712 Chr.1: 203219641-203219641 on Build GRCh38
rs770360286 Chr.1: 203219732-203219732 on Build GRCh38
rs775305615 Chr.1: 203219819-203219819 on Build GRCh38
rs148451620 Chr.1: 203219664-203219664 on Build GRCh38
rs1047175658 Chr.1: 203219719-203219719 on Build GRCh38
rs756524520 Chr.1: 203219782-203219782 on Build GRCh38
rs764074528 Chr.1: 203219773-203219773 on Build GRCh38
rs761717527 Chr.1: 203219828-203219828 on Build GRCh38
rs145366738 Chr.1: 203219799-203219799 on Build GRCh38
rs775467175 Chr.1: 203219746-203219746 on Build GRCh38
rs560698644 Chr.1: 203219772-203219772 on Build GRCh38
rs779311123 Chr.1: 203219678-203219678 on Build GRCh38
rs202073190 Chr.1: 203219682-203219682 on Build GRCh38
rs750476665 Chr.1: 203219667-203219667 on Build GRCh38
rs140151000 Chr.1: 203219777-203219777 on Build GRCh38
rs765552919 Chr.1: 203219786-203219786 on Build GRCh38
rs745573331 Chr.1: 203219741-203219741 on Build GRCh38
rs61745299 Chr.1: 203219815-203219815 on Build GRCh38
rs756171525 Chr.1: 203219670-203219670 on Build GRCh38
rs748630131 Chr.1: 203219679-203219679 on Build GRCh38
rs753015984 Chr.1: 203219789-203219789 on Build GRCh38
rs755334338 Chr.1: 203219674-203219674 on Build GRCh38
rs752675073 Chr.1: 203219647-203219647 on Build GRCh38
rs768580438 Chr.1: 203219824-203219824 on Build GRCh38
rs200338489 Chr.1: 203219778-203219778 on Build GRCh38
rs778285508 Chr.1: 203219800-203219800 on Build GRCh38
rs747474187 Chr.1: 203219801-203219801 on Build GRCh38
rs774169158 Chr.1: 203219774-203219774 on Build GRCh38
rs372798323 Chr.1: 203219810-203219810 on Build GRCh38
rs754015184 Chr.1: 203219673-203219673 on Build GRCh38
rs144580636 Chr.1: 203219742-203219742 on Build GRCh38
rs755240120 Chr.1: 203219785-203219785 on Build GRCh38
rs369555692 Chr.1: 203219638-203219638 on Build GRCh38
rs182143561 Chr.1: 203219812-203219812 on Build GRCh38
rs138767766 Chr.1: 203219820-203219820 on Build GRCh38
rs150640686 Chr.1: 203219715-203219715 on Build GRCh38
rs763035847 Chr.1: 203219653-203219653 on Build GRCh38
rs780269817 Chr.1: 203219671-203219671 on Build GRCh38
rs766463084 Chr.1: 203219847-203219847 on Build GRCh38
rs765127081 Chr.1: 203219646-203219646 on Build GRCh38
rs776279541 Chr.1: 203219739-203219739 on Build GRCh38
rs546717884 Chr.1: 203219727-203219727 on Build GRCh38
rs143518872 Chr.1: 203219814-203219814 on Build GRCh38

View Full Product Details