Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44049961-44050226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCSL
Entrez Gene ID
390110
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
Gene Aliases
-
UniGene
Hs.558851
Gene Chromosome Location
Chr.11: 43920987-44060667 on Build GRCh38
Transcripts
NM_001031854.2 XM_017017714.1 XM_017017715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6577455 c.529G>A p.E177K Chr.11: 44050086-44050086 on Build GRCh38
7175947 c.559G>A p.E187K Chr.11: 44050116-44050116 on Build GRCh38
5360764 c.558T>C p.T186T Chr.11: 44050115-44050115 on Build GRCh38
926930 c.563G>T p.R188I Chr.11: 44050120-44050120 on Build GRCh38
5448174 c.564+10T>G p.? Chr.11: 44050131-44050131 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778299756 Chr.11: 44049991-44049991 on Build GRCh38
rs757771595 Chr.11: 44049987-44049987 on Build GRCh38
rs186007139 Chr.11: 44050179-44050179 on Build GRCh38
rs775535182 Chr.11: 44050159-44050159 on Build GRCh38
rs772178609 Chr.11: 44050155-44050155 on Build GRCh38
rs374307818 Chr.11: 44050011-44050011 on Build GRCh38
rs746728486 Chr.11: 44050111-44050111 on Build GRCh38
rs767748158 Chr.11: 44050019-44050019 on Build GRCh38
rs774295748 Chr.11: 44050014-44050014 on Build GRCh38
rs765736171 Chr.11: 44050077-44050077 on Build GRCh38
rs564618022 Chr.11: 44050198-44050198 on Build GRCh38
rs760902868 Chr.11: 44050170-44050170 on Build GRCh38
rs768615056 Chr.11: 44050006-44050006 on Build GRCh38
rs758036173 Chr.11: 44050101-44050101 on Build GRCh38
rs965513873 Chr.11: 44050117-44050117 on Build GRCh38
rs908570987 Chr.11: 44050199-44050199 on Build GRCh38
rs11037842 Chr.11: 44050027-44050027 on Build GRCh38
rs764310046 Chr.11: 44050172-44050172 on Build GRCh38
rs776890973 Chr.11: 44050173-44050173 on Build GRCh38
rs779808485 Chr.11: 44050005-44050005 on Build GRCh38
rs560305489 Chr.11: 44050178-44050178 on Build GRCh38
rs34051395 Chr.11: 44050177-44050177 on Build GRCh38
rs775737201 Chr.11: 44050021-44050021 on Build GRCh38
rs779681969 Chr.11: 44050108-44050108 on Build GRCh38
rs759570626 Chr.11: 44050017-44050017 on Build GRCh38
rs745318674 Chr.11: 44049998-44049998 on Build GRCh38
rs754176104 Chr.11: 44050042-44050042 on Build GRCh38
rs367848267 Chr.11: 44050106-44050106 on Build GRCh38
rs771886008 Chr.11: 44050004-44050004 on Build GRCh38
rs757709839 Chr.11: 44050056-44050056 on Build GRCh38
rs371168564 Chr.11: 44050035-44050035 on Build GRCh38
rs747905889 Chr.11: 44050135-44050135 on Build GRCh38
rs746180065 Chr.11: 44050130-44050131 on Build GRCh38
rs374250575 Chr.11: 44050068-44050068 on Build GRCh38
rs781766904 Chr.11: 44050046-44050049 on Build GRCh38
rs80110507 Chr.11: 44050193-44050193 on Build GRCh38
rs749199530 Chr.11: 44050147-44050147 on Build GRCh38
rs749927003 Chr.11: 44050100-44050100 on Build GRCh38
rs956924684 Chr.11: 44050073-44050073 on Build GRCh38
rs754661549 Chr.11: 44050115-44050115 on Build GRCh38
rs200625908 Chr.11: 44050143-44050143 on Build GRCh38
rs776478469 Chr.11: 44050007-44050007 on Build GRCh38
rs780860183 Chr.11: 44050123-44050123 on Build GRCh38
rs769647210 Chr.11: 44050137-44050137 on Build GRCh38
rs748091293 Chr.11: 44050010-44050010 on Build GRCh38
rs988421675 Chr.11: 44050095-44050095 on Build GRCh38

View Full Product Details